Skip to Content
Merck
All Photos(1)

Documents

T3952

Sigma-Aldrich

Anti-β-Tubulin III antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Tuj 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBB3(10381)
mouse ... Tubb3(22152)
rat ... Tubb3(246118)

Looking for similar products? Visit Product Comparison Guide

General description

Microtubules are heterodimeric proteins that comprise of α- and β-tubulins. β-tubulins have several different isotypes . β-Tubulin III is a marker for nerve cell differentiation, as well as for venous and lymphatic valves . Changes in β-tubulin III expression have been associated with medulloblastoma
Anti-tubulin antibodies are useful to study the localization of tubulin proteins. Anti-β-Tubulin III antibody is specific for β-Tubulin III in humans.
Tubulin β 3 class III (TUBB3) is mapped to human chromosome 16q24.3.

Immunogen

synthetic peptide corresponding to residues 151-165 of human β-Tubulin III.

Application

Anti-β-Tubulin III antibody is suitable for use in flow cytometry (1:200-1:500), immunohistochemistry (1:100-1:200), immunoblot (approx. 50kDa) and western blot (1:500-1:1,000).
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)
Immunofluorescence (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

Missense mutations in the tubulin β 3 class III gene (TUBB3) impacts microtubule organisation in neuronal cells and contributes to neurological disorders. Mutations in TUBB3 is implicated in tubulopathies contributing to malformations in developing fetus.

Physical form

solution in phosphate buffered saline, containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutations in tubulin genes are frequent causes of various foetal malformations of cortical development including microlissencephaly.
Fallet-Bianco C, et al.
Acta Neuropathologica Communications, 2(1), 69-69 (2014)
Valentina Caracciolo et al.
Journal of cellular physiology, 223(2), 519-529 (2010-02-18)
In previous studies, we have shown overexpression and ectopic subcellular distribution of gamma-tubulin and betaIII-tubulin in human glioblastomas and glioblastoma cell lines (Katsetos et al., 2006, J Neuropathol Exp Neurol 65:455-467; Katsetos et al., 2007, Neurochem Res 32:1387-1398). Here we
Prognostic utility of beta-tubulin isotype III and correlations with other molecular and clinicopathological variables in patients with early breast cancer: a translational Hellenic Cooperative Oncology Group (HeCOG) study.
Pentheroudakis G, et al.
Breast Cancer Research and Treatment, 127(1), 179-193 (2011)
Expression of neuron-specific β-III tubulin during olfactory neurogenesis in the embryonic and adult rat.
Roskams, A.,J., et al.
Neuroscience, 83(1), 191-200 (199)
Mutations in the neuronal beta-tubulin subunit TUBB3 result in malformation of cortical development and neuronal migration defects.
Poirier K, et al.
Human Molecular Genetics, 19(22), 4462-4473 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service