Skip to Content
Merck
All Photos(1)

Key Documents

T104

Sigma-Aldrich

Tropisetron monohydrochloride

solid, ≥98% (HPLC)

Synonym(s):

1H-Indole-3-carboxylic acid (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester monohydrochloride, 3-Tropanylindole-3-carboxylate monohydrochloride, ICS-205,930, Navoban

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2O2 · HCl
CAS Number:
Molecular Weight:
320.81
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

H2O: >10 mg/mL

originator

Novartis

storage temp.

2-8°C

SMILES string

Cl[H].CN1C2CCC1CC(C2)OC(=O)c3c[nH]c4ccccc34

InChI

1S/C17H20N2O2.ClH/c1-19-11-6-7-12(19)9-13(8-11)21-17(20)15-10-18-16-5-3-2-4-14(15)16;/h2-5,10-13,18H,6-9H2,1H3;1H

InChI key

XIEGSJAEZIGKSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tropisetron monohydrochloride was used as a standard to determine the tropisetron in human plasma using HPLC method with UV detection.

Biochem/physiol Actions

3-Tropanylindole-3-carboxylate hydrochloride is a selective 5-HT3 serotonin receptor antagonist.
Selective 5-HT3 serotonin receptor antagonist.
Tropisetron a selective serotonin 5-HT3 receptor antagonist is used mainly as an antiemetic to treat nausea and vomiting following chemotherapy . In the treatment of fibromyalgia syndrome (FMS) tropisetron reduced not only pain perception but also had a favourable effect on cardiac dysfunction during treatment.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Yoshida et al.
The Journal of pharmacology and experimental therapeutics, 256(1), 272-278 (1991-01-01)
We studied the effect of 5-hydroxytryptamine 3 (5-HT3) receptor antagonists on gastrointestinal (GI) motor activity in conscious dogs with force transducers implanted chronically. During the digestive state, GR38032F, BRL 43694 and ICS 205-930 did not affect GI motor activity at
E Beubler et al.
European journal of pharmacology, 248(2), 157-162 (1993-08-02)
The effects of cholera toxin and heat stable Escherichia coli (E. coli) enterotoxin on intestinal fluid secretion are commonly considered to be mediated by cyclic nucleotides. It was demonstrated recently, by using the 5-hydroxytryptamine (5-HT)2 receptor antagonist ketanserin and the
Li Yu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 846(1-2), 20-23 (2006-09-12)
A simple and sensitive high performance liquid chromatography method with UV detection was described for the determination of tropisetron in human plasma. The prepared sample solution was injected onto BDS-C(8) reversed column using a mixture of ammonium acetate (100 mM
Q Pei et al.
European journal of pharmacology, 230(1), 63-68 (1993-01-05)
The effects of three different 5HT3 receptor antagonists, granisetron, ICS 205-930 and ondansetron (0.01, 0.1 and 1 mg/kg, s.c.) were tested on changes in mesolimbic dopamine function produced by 1 mg/kg of morphine in the rat. Increases of in vivo
Orie Tomoyose et al.
Journal of pharmacological sciences, 126(2), 136-145 (2014-09-26)
Fluvoxamine, a selective serotonin (5-HT) reuptake inhibitor, has been shown to exert analgesic effects in humans and laboratory animals. However, its effects on spinal nociceptive synaptic transmission have not been fully characterized. Here, whole-cell recordings were made from dorsal horn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service