Skip to Content
Merck
All Photos(1)

Documents

T0951

Sigma-Aldrich

Anti-Tet Repressor (TETR) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Tetracycline Repressor Protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~22 kDa by calculation

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:1,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

Tet Repressor (TETR) is homodimer comprising a C-terminal helix-turn-helix motif (HTH) and N-terminal DNA-binding domain,. It also has a C-terminal tetracycline-binding and dimerization (TBD) domain.

Immunogen

Anti-Tet Repressor (TETR) reacts with amino acid residues 31-49 (AQKLGVEQPTLYWHVKNKR) of 22 kDa Tet repressor protein

Application

Anti-Tet Repressor (TETR) antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

The Tet system is employed in conditional gene expression studies in eukaryotic cells. TetR ( can be used for such applications in some organisms without further modifications. In mammals and in a large variety of other organisms, however, eukaryotic transcriptional activator or repressor domains are fused to TetR to turn it into an efficient regulator. TetR interaction with antibiotic tetracyclin decreases its binding affinity to DNA.

Physical form

Solution of 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The induction of folding cooperativity by ligand binding drives the allosteric response of tetracycline repressor
Reichheld SE, et al.
Proceedings of the National Academy of Sciences of the USA, 106(52), 22263-22268 (2009)
The application of Tet repressor in prokaryotic gene regulation and expression
Bertram R and Hillen W
Microbial Biotechnology, 1(1), 2-16 (2008)
FOXO1 is an essential regulator of pluripotency in human embryonic stem cells
Zhang X, et al.
Nature Cell Biology, 13(9), 1092-1092 (2011)
The TetR family of transcriptional repressors
Ramos JL, et al.
Microbiology and Molecular Biology Reviews, 69(2), 326-356 (2005)
Alicia Broto et al.
Nature communications, 13(1), 1910-1910 (2022-04-09)
Mycoplasmas have exceptionally streamlined genomes and are strongly adapted to their many hosts, which provide them with essential nutrients. Owing to their relative genomic simplicity, Mycoplasmas have been used to develop chassis for biotechnological applications. However, the dearth of robust

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service