Skip to Content
Merck
All Photos(1)

Documents

T0442

Sigma-Aldrich

Anti-phospho-Tyk2 (pTyr1054/1055) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

134 kDa

species reactivity

human

technique(s)

western blot: 1:1000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TYK2(7297)

General description

Tyk2 is a member of the Jak family of protein tyrosine kinases. It associates with and is activated by
receptors for many cytokines including IL-13, the IL-6 family, IL-10, and IFN-a and β. Following ligand binding,
Tyk2 is activated by phosphorylation of Tyr1054 and/or Tyr1055. Tyk2 is required for the tyrosine phosphoryla-
tion of Stat3 in the IFN-β signaling cascade.

Specificity

Detects human Tyk2 only when phosphorylated at Tyr1054/1055.

Immunogen

synthetic phospho-Tyr1054/1055 peptide corresponding to residues around Tyr1054/1055 of human Tyk2.

Physical form

Solution in 10 mM sodium HEPES, pH 7.5, containing 150 mM sodium chloride, 100 μg/ml bovine serum albumin and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service