SYNF819200SN
Synthware™ Volumetric flask, Class A with PFTE stopper
borosilicate glass 3.3, capacity 200 mL
Synonym(s):
Synthware F819200SN, measuring flask, graduated flask
About This Item
Recommended Products
material
PTFE
borosilicate glass 3.3
feature
class A
packaging
pack of 1 ea
manufacturer/tradename
Kemtech F819200SN
capacity
200 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service