Skip to Content
Merck
All Photos(1)

Key Documents

SRP6053

Sigma-Aldrich

ProBDNF human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

Abrineurin, BDNF, Brain-derived neurotrophic factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

~52 kDa

packaging

pkg of 10 μg

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... BDNF(627)

General description

Brain-derived neurotrophic factor (BDNF) is a member of the neurotrophin family of growth factors. The precursor form of brain-derived neurotrophic factor (pro-BDNF) undergoes post-translational cleavage to form mature BDNF (mBDNF). The gene encoding this protein is localized on human chromosome 11p14.1.

Biochem/physiol Actions

Precursor form of brain-derived neurotrophic factor (pro-BDNF) interacts preferentially with the pan-neurotrophin receptor p75 (p75NTR) (2) and vps10p domain-containing receptor sortilin and induces neuronal apoptosis, whereas mature BDNF selectively binds with high affinity to the tropomyosin-related kinase B (TrkB) kinase receptor and promotes the survival, growth and differentiation of neurons. As proneurotrophins and mature neurotrophins elicit opposite biological effects, pro-BDNF cleavage in the neuronal system is regulated in a specific and cell-context dependent manner. Pro-BDNF plays important role in negative regulation of neurotrophic actions in the brain.

Physical form

Lyophilized from a 0.2 μm filtered solution of 20 mM PB and 250 mM NaCl, pH 7.2.

Preparation Note

Centrifuge the vial prior to opening.

Reconstitution

Dissolve in 1x PBS (It is not recommended to reconstitute to a final concentration less than 100 μg/mL.). This can further be diluted to other aqueous buffers.

Other Notes

Recombinant Human precursor form of Brain-derived neurotrophic factor produced in E. coli is a non-glycosylated polypeptide chain containing 2X229 amino acids as a homodimer and having a molecular mass of 52 kDa. The sequence of the first five N-terminal amino acids was determined and was found to be Ala-Pro-Met-Lys-Glu.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Function and evolution in the NGF family and its receptors.
Ebendal T
Journal of Neuroscience Research, 32(4), 461-470 (1992)
11p14.1 microdeletions associated with ADHD, autism, developmental delay, and obesity.
Shinawi M
American Journal of Medical Genetics. Part A, 155A(6), 1272-1280 (2011)
The presence of a single-nucleotide polymorphism in the BDNF gene affects the rate of locomotor adaptation after stroke.
Helm EE
Experimental Brain Research. Experimentelle Hirnforschung. Experimentation Cerebrale, 234(2), 341-351 (2016)
Hao-Yun Zhang et al.
Biochimica et biophysica acta, 1843(12), 2967-2975 (2014-09-23)
Iron accumulation is observed in the substantia nigra of patients with Parkinson's disease. However, it is unknown whether neurotrophic factors, brain-derived neurotrophic factor (BDNF) and glial cell line-derived neurotrophic factor (GDNF) participate in the modulation of neuronal iron metabolism. Here
Guoqi Zhu et al.
British journal of pharmacology, 172(9), 2354-2368 (2015-01-07)
Mild cognitive deficit in early Parkinson's disease (PD) has been widely studied. Here we have examined the effects of memantine in preventing memory deficit in experimental PD models and elucidated some of the underlying mechanisms. I.p. injection of 1-methyl-4- phenyl-1,2,3,6-tetrahydro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service