Skip to Content
Merck
All Photos(1)

Key Documents

SRP4145

Sigma-Aldrich

IL-6 from rat

recombinant, expressed in E. coli, ≥90% (SDS-PAGE), ≥90% (HPLC)

Synonym(s):

B cell differentiation factor (BCDF), BSF-2, HPGF, HSF, IFN-b2, MGI-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

rat

recombinant

expressed in E. coli

Assay

≥90% (HPLC)
≥90% (SDS-PAGE)

form

lyophilized

potency

<0.01 ng/mL ED50

mol wt

~21.7 kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Il6(24498)

General description

Interleukin- 6 (IL-6) is a potent lymphoid cell growth factor that stimulates the growth and survivability of certain B cells and T cells. It is a pro-inflammatory cytokine. IL-6 originates from mononuclear phagocytes, T and B lymphocytes, vascular endothelial cells and fibroblasts. It has a role in various physiological and pathological processes. IL-6 plays an important role in intracellular signaling pathways.[1][2] Recombinant rat IL-6 is a 21.7kDa protein containing 187 amino acid residues.

Physical form

Lyophilized without any additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in 10 mM HCl to a concentration of 0.1-1.0 μg/μL. The solution can then be diluted into other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meta-analysis of the role of IL-6 rs1800795 polymorphism in the susceptibility to prostate cancer: Evidence based on 17 studies.
Liu TZ
Medicine (2017)
Yongpan Huang et al.
BMC complementary medicine and therapies, 23(1), 2-2 (2023-01-04)
The necessity of increasing the efficiency of organ preservation has encouraged researchers to explore the mechanisms underlying diabetes-related myocardial injuries. This study intended to evaluate the protective effects of oxymatrine (OMT) in myocardial injury caused by type 2 diabetes mellitus.
Targeting interlukin-6 to relieve immunosuppression in tumor microenvironment.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine (2017)
Elin Banke et al.
Metabolism: clinical and experimental, 63(6), 831-840 (2014-04-02)
The bacteria Staphylococcus aureus is part of the normal bacterial flora and produces a repertoire of enterotoxins which can cause food poisoning and toxic shock and might contribute to the pathogenesis of inflammatory diseases. These enterotoxins directly cross-link the T
Lorena M Amaral et al.
American journal of obstetrics and gynecology, 211(2), 158-158 (2014-02-20)
Preeclampsia is a multisystem disorder recognized as hypertension with proteinuria developing >20 weeks' gestation. Preeclampsia is associated with chronic immune activation characterized by increased T and B lymphocytes, cytokines, and antibodies activating the angiotensin II type I receptor (AT1-AA). Hypertension

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service