Skip to Content
Merck
All Photos(1)

Documents

SRP2091

Sigma-Aldrich

FGF-1 human

recombinant, expressed in insect cells, ≥85% (SDS-PAGE)

Synonym(s):

AFGF, ECGF, ECGF-beta, ECGFA, ECGFB, FGF-alpha, FGFA, GLIO703, HBGF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

biological source

human

recombinant

expressed in insect cells

Assay

≥85% (SDS-PAGE)

form

frozen liquid

mol wt

~19.2 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

500 μg/mL

color

colorless to clear

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FGF1(2246)

General description

The gene FGF1 (fibroblast growth factor 1) encodes a member of the fibroblast growth factor (FGF) family. It encodes a pro-angiogenic protein that is ubiquitously expressed and is secreted via transmembrane translocation from the cells through a nonclassical secretory pathway in response to stress. It has the ability to permeate membranes composed of acidic phospholipids.

Application

FGF-1 (fibroblast growth factor 1) has been used as a growth factor to study the effect of FGF1 on early branching events in isolated endoderm obtained from embryonic mouse lung. It has also been used to study the neuroprotection effect of FGF1 on cerebellar granule neurons obtained from rats.

Biochem/physiol Actions

Acidic fibroblast growth factor (FGF-1) and basic fibroblast growth factor (FGF-2) are ubiquitous cytokines found in many tissues. They have effects on multiple cell types derived from mesoderm and neuroectoderm, including endothelial cells. FGF proteins are small peptides of 155 to 268 amino acid residues. The degree of sequence identity between different family members is 30-60 % in a "central domain" of approx. 120 amino acids. This domain confers to FGFs a common tertiary structure and the ability to bind to heparin. Secreted FGFs signal to target cells by binding and activating cell-surface tyrosine kinase FGF receptors (FGFRs; 6, 7). The function of FGFs and FGFRs during embryonic development and adult physiology has been addressed by gain- and loss-of-function experiments in several animal model organisms. These studies have shown that FGFs act as key regulators of developmental events.
The protein fibroblast growth factor 1, functions intracellularly to protect cells against stress. The protein functions as an angiogenic factor that participates in tissue repair, carcinogenesis, and maintenance of vasculature stability. Its expression is found to be upregulated in in patients with rheumatic arthritis. It serves as a prognostic biomarker in several cancers and a potential therapeutic target for patients with non-small cell lung cancer.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Folding of Fibroblast Growth Factor 1 Is Critical for Its Nonclassical Release.
Prudovsky I
Biochemistry, 55, 1159-1167 (2016)
Upregulation of fibroblast growth factor 1 in the synovial membranes of patients with late stage osteoarthritis.
Li R
Genetics and molecular research : GMR, 14, 11191-11199 (2015)
Juan Li et al.
Human pathology, 46(12), 1821-1828 (2015-09-24)
Fibroblast growth factor (FGF) 1 is identified as a candidate cancer biomarker in several kinds of cancer. However, little is known about its expression and function in non-small cell lung cancer (NSCLC). The aim of this study is to identify
Pawel A Kolodziejski et al.
General and comparative endocrinology, 294, 113498-113498 (2020-05-04)
Fibroblast growth factor 1 (FGF-1), also known as acidic fibroblast growth factor (aFGF), is a growth factor and signaling protein encoded by the Fgf1 gene. Previous studies have shown that FGF-1 may also participate in the regulation of glucose metabolism
P Cuevas et al.
Neurological research, 17(5), 396-399 (1995-10-01)
The death of spinal axotomised motoneurons provides a useful tool for studying neurotrophic factors which could prevent motoneuron loss in vivo. Median and ulnar nerves of newborn rats were unilaterally sectioned and topically treated with either a vehicle solution or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service