Skip to Content
Merck
All Photos(2)

Key Documents

SRP0311

Sigma-Aldrich

HDAC6 H611A human

recombinant, expressed in baculovirus infected Sf9 cells, ≥79% (SDS-PAGE)

Synonym(s):

HD6, JM21, histone deacetylase 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥79% (SDS-PAGE)

form

aqueous solution

mol wt

161 kDa

packaging

pkg of 50 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HDAC6(10013)

General description

HDAC6 (histone deacetylase 6) belongs to the HDAC family of proteins. HDACs are responsible for deacetylation of nuclear histone and nonhistone proteins, including transcription factors. The mutation H611A results in catalytically inactive HDAC6.
Human HDAC6 with H611A mutation (GenBank Accession No. NM_006044), full length with N-terminal GST tag, MW= 161kDa, expressed in a Baculovirus expression system.

Biochem/physiol Actions

HDAC6 (histone deacetylase 6) is involved in the control of microtubules, growth factor-mediated chemotaxis, stress response in presence of misfolded protein and tumor invasion. It also participates in EGF (epidermal growth factor)-mediated β-catenin nuclear presence and activation of c-myc. In mouse model, HDAC6 is implicated in oncogene-induced tumorigenesis. HDAC6 is the main deacetylase for α-tubulin and thus, is involved in cell motility. It is also involved in the formation of SGs (stress granules) and SG proteins. Mutations in the 3′-UTR of the HDAC6 gene suppresses hsa-miR-433-mediated post-transcriptional regulation. This results in overexpression of HDAC6, thereby causing X-linked chondrodysplasia.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiaqi Liu et al.
Journal of translational medicine, 14, 7-7 (2016-01-10)
Histone deacetylase (HDAC) inhibitors are widely used in clinical investigation as novel drug targets. For example, panobinostat and vorinostat have been used to treat patients with melanoma. However, HDAC inhibitors are small-molecule compounds without a specific target, and their mechanism
Histone deacetylase 6 binds polyubiquitin through its zinc finger (PAZ domain) and copurifies with deubiquitinating enzymes.
Hook SS, et al.
Proceedings of the National Academy of Sciences of the USA, 99, 13425-13425 (2002)
Loss of a-Tubulin Acetylation Is Associated with TGF-?-induced Epithelial-Mesenchymal Transition.
Gu S, et al.
The Journal of Biological Chemistry, 291, 5396-5396 (2016)
Li S. et al.
Neurology, 41, 112-116 (2010)
HDAC11 is a novel drug target in carcinomas.
Deubzer HE, et al.
International Journal of Cancer. Journal International Du Cancer, 132, 2200-2200 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service