Skip to Content
Merck
All Photos(1)

Key Documents

SML3724

Sigma-Aldrich

Troxerutin

≥97% (HPLC)

Synonym(s):

2-[3,4-bis(2-Hydroxyethoxy)phenyl]-3-[[6-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-5-hydroxy-7-(2-hydroxyethoxy)-4H-1-benzopyran-4-one, 7,3′,4′-tris[O-(2-hydroxyethyl)]rutin, Trihydroxyethylrutin, Troxerutinum, tri(hydroxyethyl)rutoside, trioxyethylrutin, vitaminP4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H42O19
CAS Number:
Molecular Weight:
742.68
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O1[C@H]([C@@H]([C@@H]([C@H]([C@@H]1C)O)O)O)OC[C@H]2O[C@H]([C@@H]([C@H]([C@@H]2O)O)O)Oc3[c](c4c([o]c3c5cc(c(cc5)OCCO)OCCO)cc(cc4O)OCCO)=O

InChI

1S/C33H42O19/c1-14-23(38)26(41)28(43)32(49-14)48-13-21-24(39)27(42)29(44)33(51-21)52-31-25(40)22-17(37)11-16(45-7-4-34)12-20(22)50-30(31)15-2-3-18(46-8-5-35)19(10-15)47-9-6-36/h2-3,10-12,14,21,23-24,26-29,32-39,41-44H,4-9,13H2,1H3/t14-,21+,23-,24+,26+,27-

InChI key

IYVFNTXFRYQLRP-VVSTWUKXSA-N

Biochem/physiol Actions

Brain penetrant, anti-oxidative, anti-radiation, and anti-inflammatory flavonol
Troxerutin is a brain penetrant, vasoprotective flavonol isolated from Sophora japonica, which is present in many other plants including grains, fruits, and vegetables. Troxerutin exhibits multiple biological activities including anti-oxidative, anti-radiation, and anti-inflammatory. Recent study indicates that troxerutin improves the synaptic failure induced by Aβ peptide. It appears that Troxerutin could promote radioprotection by stimulating NEAT1 to upregulate PDPK1 expression by suppressing miR-147.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong-Jian Hu et al.
iScience, 26(2), 105932-105932 (2023-01-27)
A better understanding of the molecular mechanism involving the lncRNA-miRNA-mRNA network underlying radiation damage can be beneficial for radioprotection. This study was designed to investigate the potential role of lncRNA NEAT1, miR-147 and Phosphoinositide Dependent Protein Kinase 1 (PDPK1) interaction
F Fahmideh et al.
Frontiers in pharmacology, 13, 951833-951833 (2022-09-02)
Diabetic retinopathy (DR), one of the most common complications of diabetes mellitus, is characterized by degeneration of retinal neurons and neoangiogenesis. Until today, the pharmacological approaches for DR are limited and focused on counteracting the end-stage of this neurodegenerative disease
Shirin Babri et al.
European journal of pharmacology, 732, 19-25 (2014-04-01)
Alzheimer׳s disease (AD) is a neurodegenerative disorder with a progressive cognitive decline and memory loss. Multiple pathogenetic factors including aggregated β-amyloid (Aβ), neurofibrillary tangles (NFTs), cholinergic dysfunction and oxidative stress are involved in AD. Aβ, a major constituent of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service