Skip to Content
Merck
All Photos(1)

Key Documents

SML3531

Sigma-Aldrich

UPF-648

≥98% (HPLC)

Synonym(s):

(1S,2S)-2-(3,4-Dichlorobenzoyl)cyclopropanecarboxylic acid, (1S,2S)-2-[(3,4-Dichlorophenyl)carbonyl]cyclopropane-1-carboxylic acid, DBCC, UPF 648, UPF648

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8Cl2O3
CAS Number:
Molecular Weight:
259.09
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(O)[C@@H]1[C@H](C1)C(C2=CC=C(C(Cl)=C2)Cl)=O

InChI

1S/C11H8Cl2O3/c12-8-2-1-5(3-9(8)13)10(14)6-4-7(6)11(15)16/h1-3,6-7H,4H2,(H,15,16)/t6-,7-/m0/s1

InChI key

ZBRKMOHDGFGXLN-BQBZGAKWSA-N

Biochem/physiol Actions

Potent, active site-targeting kynurenine 3-monooxygenase (KMO; kynurenine 3-hydroxylase) inhibitor that protects against neurodegeneration in vivo.
UPF-648 is a potent, active site-targeting kynurenine 3-monooxygenase (KMO; kynurenine 3-hydroxylase) inhibitor (IC50 = 20 nM) that prevents productive binding of the substrate L-kynurenine by perturbing the local active-site structure. UPF-648 protects against neurodegeneration in a murine (30 mg/kg, i.p.) and a Drosophila (100 μM in maize media) model of Huntington′s disease by shifting kynurenine pathway metabolism towards enhanced neuroprotective kynurenic acid (KYNA) formation and away from the free radicals generator 3-hydroxykynurenine (3-HK) and the excitotoxic quinolinic acid (QUIN).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endogenous kynurenate controls the vulnerability of striatal neurons to quinolinate: Implications for Huntington's disease
Sapko MT, Guidetti P, Yu P, Tagle DA, Pellicciari R, Schwarcz R
Experimental Neurology, 197, 31-40 (2006)
Biochemistry and structural studies of kynurenine 3-monooxygenase reveal allosteric inhibition by Ro 61-8048
Faseb Journal, 32, 2036-2045 (2018)
Susanna Campesan et al.
Current biology : CB, 21(11), 961-966 (2011-06-04)
Neuroactive metabolites of the kynurenine pathway (KP) of tryptophan degradation have been implicated in the pathophysiology of neurodegenerative disorders, including Huntington's disease (HD) [1]. A central hallmark of HD is neurodegeneration caused by a polyglutamine expansion in the huntingtin (htt)
Tzu-Ting Huang et al.
EBioMedicine, 54, 102717-102717 (2020-04-09)
Triple-negative breast cancer (TNBC) is aggressive and has a poor prognosis. Kynurenine 3-monooxygenase (KMO), a crucial kynurenine metabolic enzyme, is involved in inflammation, immune response and tumorigenesis. We aimed to study the role of KMO in TNBC. KMO alteration and
Ewelina Rojewska et al.
Frontiers in pharmacology, 9, 724-724 (2018-07-28)
Neuropathic pain caused by a primary injury or dysfunction in the peripheral or central nervous system is a tremendous therapeutic challenge. Here, we have collected the first evidence from a single study on the potential contributions to neuropathic pain development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service