Skip to Content
Merck
All Photos(1)

Documents

SML3427

Sigma-Aldrich

C4-HSL

≥95% (HPLC)

Synonym(s):

(S)-N-(2-Oxotetrahydrofuran-3-yl)butyramide, BHL, N-Butanoyl-L-homoserine lactone, N-Butyrylhomoserine lactone, N-[(3S)-Tetrahydro-2-oxo-3-furanyl]butanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13NO3
CAS Number:
Molecular Weight:
171.19
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CCCC(N[C@@H]1C(OCC1)=O)=O

InChI

1S/C8H13NO3/c1-2-3-7(10)9-6-4-5-12-8(6)11/h6H,2-5H2,1H3,(H,9,10)/t6-/m0/s1

InChI key

VFFNZZXXTGXBOG-LURJTMIESA-N

Biochem/physiol Actions

C4-HSL (N-butyryl-L-Homoserine lactone) is a quorum-sensing signaling molecule produced by Pseudomonas aeruginosa. C4-HSL induces expression of the virulence genes lasB and rhlA in P. aeruginosa when used at a concentration of 10 µM. C4-HSL exhibits antibacterial activity against MDR S. aureus isolates but had no effect on A. baumannii.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service