Skip to Content
Merck
All Photos(2)

Key Documents

SML3390

Sigma-Aldrich

25CN-NBOH hydrochloride

≥98% (HPLC)

Synonym(s):

2-([2-(4-Cyano-2, 5-dimethoxyphenyl)ethylamino]methyl)phenol hydrochloride, 4-(2-(2-Hydroxybenzylamino)ethyl)-2,5-dimethoxybenzonitrile hydrochloride, NBOH-2C-CN hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20N2O3·HCl
CAS Number:
Molecular Weight:
348.82
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

25CN-NBOH is a brain-penetrant, highly potent and selective 5-HT2A receptor agonist (human 2A/rat 2C Ki = 1.3/132 nM; human 2A/2C PI turnover EC50 = 2.1/190 nM) with much reduced efficacy toward human 2B & 2C subtypes (Ki ratio: 2C/2A = 52-81 & 2B/2A = 37; Ca2+ mobilization Δlog(Rmax/EC50 ratio: 2C/2A = 30-180 & 2B/2A = 54; 2C/2A PI turnover EC50 ratio = 81-146) and other receptors/transporters/kinases. When administered in mice in vivo (1, 3, 6 mg/kg s.c.), 25CN-NBOH produces hallucinogen-like effects blockable by 5-HT2A antagonist M100907, but not 5-HT2C antagonist RS102221.
Brain-penetrant, highly potent and selective 5-HT2A receptor agonist with good in vitro & in vivo efficacy.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tobias Buchborn et al.
Frontiers in pharmacology, 9, 17-17 (2018-02-23)
The serotonin (5-HT) 2A receptor is the primary molecular target of serotonergic hallucinogens, which trigger large-scale perturbations of the cortex. Our understanding of how 5-HT2A activation may cause the effects of hallucinogens has been hampered by the receptor unselectivity of
William E Fantegrossi et al.
Psychopharmacology, 232(6), 1039-1047 (2014-09-17)
2-([2-(4-cyano-2,5-dimethoxyphenyl)ethylamino]methyl)phenol (25CN-NBOH) is structurally similar to N-benzyl substituted phenethylamine hallucinogens currently emerging as drugs of abuse. 25CN-NBOH exhibits dramatic selectivity for 5-HT2A receptors in vitro, but has not been behaviorally characterized. 25CN-NBOH was compared to the traditional phenethylamine hallucinogen R(-)-2,5-dimethoxy-4-iodoamphetamine
Tobias Buchborn et al.
Journal of psychopharmacology (Oxford, England), 34(7), 786-794 (2020-02-13)
Serotonin 2A receptors, the molecular target of psychedelics, are expressed by neuronal and vascular cells, both of which might contribute to brain haemodynamic characteristics for the psychedelic state. Aiming for a systemic understanding of psychedelic vasoactivity, here we investigated the
Anna U Odland et al.
Neuropharmacology, 183, 107838-107838 (2019-11-07)
5-Hydroxytryptamine 2A receptor (5-HT2AR) agonist psychedelics are increasingly recognized as potentially useful treatments of psychiatric disorders, such as obsessive-compulsive disorder, depression, anxiety, and drug dependence. There is limited understanding of the way they exert their therapeutic action, but inhibition of
Adam L Halberstadt et al.
Psychopharmacology, 236(2), 821-830 (2018-11-19)
There is evidence that mGlu2/3 receptors regulate 5-HT2A signaling, interactions that have been theorized to play a role in the antipsychotic-like effects of mGlu2/3 agonists as well as the hallucinogenic effects of 5-HT2A agonists. One approach to unraveling this interaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service