Skip to Content
Merck
All Photos(1)

Key Documents

SML3100

Sigma-Aldrich

AFMT racemate

≥98% (HPLC)

Synonym(s):

α-(Fluoromethyl)-D/L-tyrosine, (R/S)-α-Fluoromethyltyrosine, (R/S)-2-Amino-3-fluoro-2-(4-hydroxybenzyl)propanoic acid, Alpha-(FluoroMethyl)-D/L-Tyrosine, D/L-AFMT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12FNO3
CAS Number:
Molecular Weight:
213.21
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

OC1=CC=C(C=C1)CC(N)(C(O)=O)CF

InChI

1S/C10H12FNO3/c11-6-10(12,9(14)15)5-7-1-3-8(13)4-2-7/h1-4,13H,5-6,12H2,(H,14,15)

InChI key

ACIQAJKTARSFHQ-UHFFFAOYSA-N

Biochem/physiol Actions

A racemic mixture of D- and L-AFMT. L-AFMT is reported to selectively inhibt against gut bacteria E. faecalis pyridoxal-5′-phosphate (PLP)-dependent tyrosine decarboxylase (TyrDC)-, but not aromatic amino acid decarboxylase (AADC)-, mediated L-dopa decarboxylation (IC50 = 4.7 μM/cell-free, 1.4 μM/in E. faecalis cultures; 20% human AADC inhibition at 650 μM) via covalent adduct formation with TyrDC co-factor PLP. When co-administered with L-dopa (10 mg/kg) and the AADC inhibitor carbidopa (30 mg/kg) to gnotobiotic mice colonized with E. faecalis, L-AFMT (25 mg/kg) significantly increases L-dopa peak serum concentration.
Covalent inhibtor against L-dopa decarboxylation by gut bacteria E. faecalis tyrosine decarboxylase (TyrDC), but not aromatic amino acid decarboxylase (AADC).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vayu Maini Rekdal et al.
Science (New York, N.Y.), 364(6445) (2019-06-15)
The human gut microbiota metabolizes the Parkinson's disease medication Levodopa (l-dopa), potentially reducing drug availability and causing side effects. However, the organisms, genes, and enzymes responsible for this activity in patients and their susceptibility to inhibition by host-targeted drugs are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service