Skip to Content
Merck
All Photos(1)

Key Documents

SML2998

Sigma-Aldrich

Moxidectin

≥98% (HPLC)

Synonym(s):

Moxidectin, (6R,23E,25S)-5-O-Demethyl-28-deoxy-25-[(1E)-1,3-dimethyl-1-buten-1-yl]-6,28-epoxy-23-(methoxyimino)milbemycin B, 23-Methoxime-LL-F 28249α, CL 301,423, CL 301423, CL301,423, CL301423, Equest, ProHeart 6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H53NO8
CAS Number:
Molecular Weight:
639.82
MDL number:
UNSPSC Code:
12352119
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CO\N=C1/C[C@]2(C[C@@H]3CC(C\C=C(C)\C[C@@H](C)\C=C\C=C4/COC5[C@H](O)C(C)=C[C@@H](C(=O)O3)[C@]45O)O2)O[C@@H]([C@H]1C)\C(C)=C\C(C)C

InChI

1S/C37H53NO8/c1-21(2)14-25(6)33-26(7)31(38-42-8)19-36(46-33)18-29-17-28(45-36)13-12-23(4)15-22(3)10-9-11-27-20-43-34-32(39)24(5)16-30(35(40)44-29)37(27,34)41/h9-12,14,16,21-22,26,28-30,32-34,39,41H,13,15,17-20H2,1-8H3/b10-9+,23-12+,25-14+,27-11+,38-31+/t22-,26-,28+,29-,30-,32+,33+,34+,36-,37+/m0/s1

InChI key

YZBLFMPOMVTDJY-LSGXYNIPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Moxidectin is an orally active antihelminthic (antiparasitic) that targets parasite glutamate-gated chloride ion channels (GluCls). Moxidectin binding increases target GluCl permeability, resulting in influx of chloride ions and dysfunction of the excretory pore, flaccid paralysis, and death of the parasite through host response.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valentin Buchter et al.
ACS infectious diseases, 7(5), 1069-1076 (2020-09-30)
Strongyloides stercoralis is a soil-transmitted helminth affecting an estimated 30-100 million people. Since the infection may be severe and life-threatening, accessible and effective treatment is pivotal. Currently, ivermectin is the drug of choice but has limitations. Moxidectin, a veterinary anthelminthic
Mahmoud Sharaf et al.
Experimental parasitology, 217, 107961-107961 (2020-08-11)
Scabies is considered one of the commonest dermatological diseases that has a global health burden. Current treatment with ivermectin (IVM) is insufficient and potential drug resistance was noticed. Moxidectin (MOX), with a better pharmacological profile may be a promising alternative.
T Paterson et al.
Veterinary parasitology, 284, 109194-109194 (2020-09-01)
Canine heartworm disease (CHD) results from infection with Dirofilaria immitis and while it is of global concern, it is most prevalent in tropical climates where conditions support the parasite and vector life cycles. Melarsomine dihydrochloride is the sole treatment for
Wieland Beck
Veterinary parasitology, 283, 109190-109190 (2020-08-02)
An outbreak of sarcoptic mange was investigated in a herd of llamas and alpacas in the Black Forrest (Baden-Wuerttemberg, Germany). The diagnosis was made by clinical picture and detection of mites in skin scrapings and ear swabs. At the beginning
Raj Packianathan et al.
Parasites & vectors, 13(1), 448-448 (2020-09-07)
The Australian paralysis tick, Ixodes holocyclus, causes tick paralysis in dogs and cats in the eastern coastal regions of Australia. Prevention is the best option to protect dogs against this potentially fatal disease and sarolaner provides rapid and sustained efficacy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service