Skip to Content
Merck
All Photos(1)

Documents

SML2947

Sigma-Aldrich

Penehyclidine hydrochloride

≥98% (HPLC)

Synonym(s):

α-[(1-Azabicyclo[2.2.2]oct-3-yloxy)methyl]-α-cyclopentyl-benzenemethanol hydrochloride, 3-(2′-phenyl-2′-cyclopentyl-2′-hydroxyl-ethoxy) quinuclidine, Penequine hydrochloride, Penequinine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H29NO2 · HCl
CAS Number:
Molecular Weight:
351.91
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder (hygroscopic)

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

InChI

1S/C20H29NO2.ClH/c22-20(18-8-4-5-9-18,17-6-2-1-3-7-17)15-23-19-14-21-12-10-16(19)11-13-21;/h1-3,6-7,16,18-19,22H,4-5,8-15H2;1H

InChI key

DDTVVMRZNVIVQM-UHFFFAOYSA-N

Biochem/physiol Actions

Penehyclidine hydrochloride is a brain penetrant anticholinergic drug that exhibits both antimuscarinic and antinicotinic activity. Penehyclidine hydrochloride does not induce muscarinic 2 receptor-associated cardiovascular side effects. Penehyclidine hydrochloride is effective in experimental models of heart, lung and brain ischemia reperfusion injury.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaguang Wang et al.
European journal of pharmacology, 839, 1-11 (2018-09-12)
Penehyclidine hydrochloride (PHC) is a new anticholinergic agent that provides protective effects in experimental models of heart and brain ischaemia as well as reperfusion (I/R) injury. In this study, we tested the hypothesis that PHC can alleviate lung ischaemia-reperfusion injury
Yaguang Wang et al.
Drug design, development and therapy, 12, 3289-3299 (2018-10-17)
Penehyclidine hydrochloride (PHC) is an anticholinergic drug manufactured in China. It is used widely in clinics as a reversal agent in cases of organic phosphorus poisoning and as a preanesthetic medication. Compared with other anticholinergic agents, PHC confers substantial advantages.
Yong-An Wang et al.
Life sciences, 78(2), 210-223 (2005-09-13)
Previous studies have paid little attention to the anticonvulsant effect of anticholinergic drugs that act on both muscarinic (M) and nicotinic (N) receptors during soman-induced seizures. Therefore, with the establishment of a soman-induced seizures model in rats, this study evaluated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service