Skip to Content
Merck
All Photos(1)

Key Documents

SML2946

Sigma-Aldrich

Biotin-aniline

≥98% (HPLC)

Synonym(s):

(3aS,4S,6aR)-N-[2-(4-Aminophenyl)ethyl]hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide, Btn-An, N-(4-Aminophenethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, N-(4-Aminophenethyl)biotinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26N4O2S
CAS Number:
Molecular Weight:
362.49
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(CCCC[C@@H]1SC[C@]([H])(N2)[C@@]1(NC2=O)[H])NCCC3=CC=C(C=C3)N

InChI

1S/C18H26N4O2S/c19-13-7-5-12(6-8-13)9-10-20-16(23)4-2-1-3-15-17-14(11-25-15)21-18(24)22-17/h5-8,14-15,17H,1-4,9-11,19H2,(H,20,23)(H2,21,22,24)/t14-,15-,17-/m0/s1

Biochem/physiol Actions

Biotin-aniline (Btn-An) is a superior substrate than biotinyl tyramide (biotin-phenol; BP) for APEX2-mediated proximity labeling of RNA (∼25-fold higher by dot blot with 0.5 mM substrate and 0.4-10 μg RNA/dot). Btn-An specifically labels mitochondrial, but not cytoplasmic, mRNAs in live HEK293T stable line expressesing APEX2 in the mitochondrial matrix (mito-APEX2).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Zhou et al.
Angewandte Chemie (International ed. in English), 58(34), 11763-11767 (2019-06-27)
The subcellular organization of biomolecules such as proteins and nucleic acids is intimately linked to their biological functions. APEX2, an engineered ascorbate peroxidase that enables proximity-dependent labeling of proteins in living cells, has emerged as a powerful tool for deciphering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service