SML2826
NVS-MLLT-1
≥98% (HPLC)
Synonym(s):
(S)-N-(2-((2-Methylpyrrolidin-1-yl)methyl)-1H-benzo[d]imidazol-5-yl)isoquinoline-6-carboxamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C23H23N5O
Molecular Weight:
385.46
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
Biochem/physiol Actions
NVP-MLLT-1 is a potent and selective inhibitor against YEATS domain (YD) of MLLT1 (ENL/YEATS1) and MLLT3 (AF9/YEATS3).
Potent and selective inhibitor against YEATS domain (YD) of MLLT1 (ENL/YEATS1) and MLLT3 (AF9/YEATS3).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service