Skip to Content
Merck
All Photos(1)

Key Documents

SML2494

Sigma-Aldrich

Pralatrexate

≥98% (HPLC)

Synonym(s):

(2S)-2-[[4-[(1RS)-1-[(2,4-diaminopteridin-6-yl)methyl]but-3-ynyl]benzoyl]amino]pentanedioic acid, 10-propargyl-10-deazaaminopterin, N-(4-{1-[(2,4-diaminopteridin-6-yl)methyl]but-3-yn-1-yl}benzoyl)-L-glutamic acid, PDX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H23N7O5
CAS Number:
Molecular Weight:
477.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL

storage temp.

−20°C

SMILES string

O=C([C@@H](NC(C1=CC=C(C(CC(N=C23)=CN=C3N=C(N=C2N)N)CC#C)C=C1)=O)CCC(O)=O)O

InChI

1S/C23H23N7O5/c1-2-3-14(10-15-11-26-20-18(27-15)19(24)29-23(25)30-20)12-4-6-13(7-5-12)21(33)28-16(22(34)35)8-9-17(31)32/h1,4-7,11,14,16H,3,8-10H2,(H,28,33)(H,31,32)(H,34,35)(H4,24,25,26,29,30)/t14?,16-/m0/s1

InChI key

OGSBUKJUDHAQEA-WMCAAGNKSA-N

Biochem/physiol Actions

Pralatrexate is an antifolate that selectively enters cancer cells expressing reduced folate carrier type 1 (RFC-1). Pralatrexate potently inhibits DHFR (dihydrofolate reductase).

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Owen A O'Connor et al.
Leukemia & lymphoma, 58(11), 2548-2557 (2017-07-26)
It has been nearly 8 years since pralatrexate became the first drug approved by the U.S. Food and Drug Administration for the treatment of relapsed or refractory peripheral T-cell lymphoma (PTCL). Like most drugs approved for a particular clinical indication
Jennifer E Amengual et al.
Blood, 131(4), 397-407 (2017-11-17)
Peripheral T-cell lymphomas (PTCL) are a group of rare malignancies characterized by chemotherapy resistance and poor prognosis. Romidepsin and pralatrexate were approved by the US Food and Drug Administration for patients with relapsed/refractory PTCL, exhibiting response rates of 25% and
Enrica Marchi et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 19(24), 6657-6661 (2013-08-24)
Folates are well known to be essential for many cellular processes, including cellular proliferation. As a consequence, antifolates, the fraudulent mimics of folic acid, have been shown to be potent therapeutic agents in many cancers. Over the past several decades

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service