Skip to Content
Merck
All Photos(1)

Key Documents

SML2166

Sigma-Aldrich

KML29

≥98% (HPLC)

Synonym(s):

1,1,1,3,3,3-Hexafluoropropan-2-yl 4-(dibenzo[d][1,3]dioxol-5-yl(hydroxy)methyl)piperidine-1-carboxylate, 4-[bis(1,3-Benzodioxol-5-yl)hydroxymethyl]-1-piperidinecarboxylic acid 2,2,2-trifluoro-1-(trifluoromethyl)ethyl ester, KML 29, KML-29

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21F6NO7
CAS Number:
Molecular Weight:
549.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)C(OC(=O)N1CCC(CC1)C(O)(c4cc5c(cc4)OCO5)c2cc3c(cc2)OCO3)C(F)(F)F

InChI

1S/C24H21F6NO7/c25-23(26,27)20(24(28,29)30)38-21(32)31-7-5-13(6-8-31)22(33,14-1-3-16-18(9-14)36-11-34-16)15-2-4-17-19(10-15)37-12-35-17/h1-4,9-10,13,20,33H,5-8,11-12H2

InChI key

SXHQLPHDBLTFPM-UHFFFAOYSA-N

Biochem/physiol Actions

KML29, an O-hexafluoroisopropyl (HFIP) carbamate analogue of JZL184. is a potent and orally active monoacylglycerol lipase (monoglyceride lipase; MAGL; MGLL; MGL) inhibitor (IC50 = 5.9/15/43 nM against human/mouse/rat MAGL) that targets MAGL active site with greatly improved selectivity (rat/mouse ABHD6 IC50 = 1.60/4.87; no inhibitory activity against human/rat/mouse fatty acid amide hydrolase (FAAH) up to 50 μM). KML29 in vivo treatment results in a selective upregulation of 2-arachidonoyl glycerol (2-AG), but not N-arachidonoyl-ethanolamine (AEA) in mice (brain Emax ∼20 mg/kg p.o. or i.p.; peripheral Emax ∼1 mg/kg p.o.) and rats (brain Emax ∼40 mg/kg i.p.). KML29 reduces inflammatory and neuropathic nociceptive behaviour in animal studies without cannabimimetic side effects seen with dual FAAH & MAGL inhibition, chronic administration, however, leads to CB1 receptor desensitization as observed with other MAGL inhibitors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B M Ignatowska-Jankowska et al.
British journal of pharmacology, 171(6), 1392-1407 (2013-07-16)
Since monoacylglycerol lipase (MAGL) has been firmly established as the predominant catabolic enzyme of the endocannabinoid 2-arachidonoylglycerol (2-AG), a great need has emerged for the development of highly selective MAGL inhibitors. Here, we tested the in vivo effects of one
Molly S Crowe et al.
British journal of pharmacology, 174(23), 4523-4539 (2017-10-01)
Gabapentin is commonly prescribed for nerve pain but may also cause dizziness, sedation and gait disturbances. Similarly, inhibition of the endogenous cannabinoid enzyme monoacylglycerol lipase (MAGL) has antinociceptive and anti-inflammatory properties but also induces sedation in mice at high doses.
Andreu Viader et al.
eLife, 5, e12345-e12345 (2016-01-19)
Metabolic specialization among major brain cell types is central to nervous system function and determined in large part by the cellular distribution of enzymes. Serine hydrolases are a diverse enzyme class that plays fundamental roles in CNS metabolism and signaling.
Sally Miller et al.
Investigative ophthalmology & visual science, 57(7), 3287-3296 (2016-06-23)
Cannabinoids, such as Δ9-THC, act through an endogenous signaling system in the vertebrate eye that reduces IOP via CB1 receptors. Endogenous cannabinoid (eCB) ligand, 2-arachidonoyl glycerol (2-AG), likewise activates CB1 and is metabolized by monoacylglycerol lipase (MAGL). We investigated ocular
Noemi Pasquarelli et al.
Neurochemistry international, 110, 14-24 (2017-08-23)
The modulation of the brain endocannabinoid system has been identified as an option to treat neurodegenerative diseases including Parkinson's disease (PD). Especially the elevation of endocannabinoid levels by inhibition of hydrolytic degradation represents a valuable approach. To evaluate whether monoacylglycerol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service