Skip to Content
Merck
All Photos(1)

Key Documents

SML1988

Sigma-Aldrich

TASIN-1 hydrochloride

≥98% (HPLC)

Synonym(s):

1′-(4-Methoxyphenylsulfonyl)-4-methyl-1,4′-bipiperidine hydrochloride, 1,4′-Bipiperidine, 1′-[(4-methoxyphenyl)sulfonyl]-4-methyl-, hydrochloride, Truncated APC Selective INhibitor-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H28N2O3S · HCl
CAS Number:
Molecular Weight:
388.95
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(CC1)CCN1C(CC2)CCN2S(C3=CC=C(OC)C=C3)(=O)=O.[H]Cl

InChI key

TXDTUQPRRXNCLP-UHFFFAOYSA-N

Biochem/physiol Actions

EBP-dependent cholesterol synthesis inhibitor with selective cytotoxicity against colorectal cancer cells harboring APC truncations.
TASIN-1 (Truncated APC Selective INhibitor-1) targets EBP- (Emopamil-binding protein; D8-D7 sterol isomerase) dependent cholesterol biosynthesis and selectively induces cellular cholesterol depletion and apoptotic death in human colorectal cancer (CRC) cultures harboring APC truncations under lipid-deficient conditions (0.2% FCS or 2% lipoprotein-deficient serum; IC50 from 35 nM/HT29 to 4.9 μM/HT55 in 72 hrs) as a result of defective cholesterol homeostasis regulation, while exhibiting little cytotoxicity toward wild-type APC CRC cells or non-transformed human colonic and bronchial epithelial cells (IC50 >50 μM in 72 hrs). TASIN-1 intraperitoneal injection is efficacious against the growth of CRC tumors with trunctated APC using the genetically engineered CPC;Apc mice (20 mg/kg twice a week) and two xenograft murine models (DLD1 & HT29; 40 mg/kg twice daily) without apparent toxicity to the animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu Zhang et al.
Science translational medicine, 8(361), 361ra140-361ra140 (2016-11-01)
Mutations in the adenomatous polyposis coli (APC) gene are common in colorectal cancer (CRC), and more than 90% of those mutations generate stable truncated gene products. We describe a chemical screen using normal human colonic epithelial cells (HCECs) and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service