Skip to Content
Merck
All Photos(1)

Key Documents

SML1939

Sigma-Aldrich

SCH 79797 hydrochloride

≥98% (HPLC)

Synonym(s):

N3-Cyclopropyl-7-(4-isopropylbenzyl)-7H-pyrrolo[3,2-f]quinazoline-1,3-diamine hydrochloride, N3-Cyclopropyl-7-[[4-(1-methylethyl)phenyl]methyl]-7H-Pyrrolo[3,2-f]quinazoline-1,3-diamine hydrochloride, SCH79797 xHCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25N5 · xHCl
CAS Number:
Molecular Weight:
371.48 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to light brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

Cl.Cl.[n]2(c3c(c4c(nc(nc4N)NC5CC5)cc3)cc2)Cc1ccc(cc1)C(C)C

InChI

1S/C23H25N5.2ClH/c1-14(2)16-5-3-15(4-6-16)13-28-12-11-18-20(28)10-9-19-21(18)22(24)27-23(26-19)25-17-7-8-17;;/h3-6,9-12,14,17H,7-8,13H2,1-2H3,(H3,24,25,26,27);2*1H

InChI key

NNJTXSQXGHYXAJ-UHFFFAOYSA-N

Biochem/physiol Actions

SCH 79797 aids protection during ischemic injury. During subarachnoid hemorrhage (SAH), it plays a key role in the maintenance of microvascular integrity. SCH 79797 also elicits pro‐apoptotic and antiproliferative effects.
SCH 79797 is a nonpeptidic antagonist that targets protease activated receptor-1 (PAR-1) with high affinity (IC50 = 70 nM against 10 nM haTRAP in competitive binding assay; Ki = 35 nM) and selectively inhibits PAR-1 agonists-induced human platelet aggregation (IC50 = 300 nM and 3 μM against 300 nM haTRAP and 0.1 U/mL α-thrombin, respectively), while exhibiting little potency against platelet aggregation induced by ADP, collagen, or PAR-4 agonist γ-thrombin. SCH-79797 is shown to display neuroprotective and antiepileptogenic efficacy in a rat model of lithium/pilocarpine-induced status epilepticus (SE) in vivo (25 μg/kg via daily i.p. 20-30 min after SE termination).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SCH 79797, a selective PAR1 antagonist, protects against ischemia/reperfusion-induced arrhythmias in the rat hearts
Mohamed OY, et al.
European Review for Medical and Pharmacological Sciences, 20(22), 4796-4800 (2016)
Protease-activated receptor 1-selective antagonist SCH79797 inhibits cell proliferation and induces apoptosis by a protease-activated receptor 1-independent mechanism
Di Serio C, et al.
Basic and Clinical Pharmacology and Toxicology, 101(1), 63-69 (2007)
Role of SCH79797 in maintaining vascular integrity in rat model of subarachnoid hemorrhage
Yan J, et al.
Stroke, 44(5), 1410-1417 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service