Skip to Content
Merck
All Photos(1)

Key Documents

SML1906

Sigma-Aldrich

AS1517499

≥98% (HPLC)

Synonym(s):

2-[[2-(3-Chloro-4-hydroxyphenyl)ethyl]amino]-4-[(phenylmethyl)amino]-5-pyrimidinecarboxamide, 4-(Benzylamino)-2-{[2-(3-chloro-4-hydroxyphenyl)ethyl]amino}pyrimidine-5-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20ClN5O2
CAS Number:
Molecular Weight:
397.86
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC(CCNC2=NC=C(C(N)=O)C(NCC3=CC=CC=C3)=N2)=CC=C1O

InChI

1S/C20H20ClN5O2/c21-16-10-13(6-7-17(16)27)8-9-23-20-25-12-15(18(22)28)19(26-20)24-11-14-4-2-1-3-5-14/h1-7,10,12,27H,8-9,11H2,(H2,22,28)(H2,23,24,25,26)

InChI key

OZRMEKAUZBKTTC-UHFFFAOYSA-N

Biochem/physiol Actions

AS1517499 is a potent STAT6 (signal transducer and activator of transcription 6) inhibitor (IC50 = 21 nM against IL-4-dependent reporter activity) that selectively prevents anti-CD3- & IL-4-induced T-helper cell 2 (Th2) differentiation of mouse spleen T cells (IC50 = 2.3 nM), without affecting anti-CD3- & IL-12-induced Th1 differentiation. AS1517499 in vivo effecicacy is demonstrated in various animal models of immunological disorders, including the autoimmune Graves′ disease, antigen-induced bronchial hypercontractility (10 mg/kg i.p.). AS1517499 treatment (10 mg/kg/day i.p.) following early gliomagenesis blockage by a CSF-1R inhibitor is also shown to greatly prolong the survival rate when compared with CSF-1R inhibition alone in a murine model of PDGF-driven glioma (PDG).
AS1517499 moderately inhibits the expression of RhoA (ras homolog family member A). Overexpression of RhoA protein is associated with allergic bronchial asthma.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela F Quail et al.
Science (New York, N.Y.), 352(6288), aad3018-aad3018 (2016-05-21)
Macrophages accumulate with glioblastoma multiforme (GBM) progression and can be targeted via inhibition of colony-stimulating factor-1 receptor (CSF-1R) to regress high-grade tumors in animal models of this cancer. However, whether and how resistance emerges in response to sustained CSF-1R blockade
The proximal STAT6 and NF-κB sites are responsible for IL-13-and TNF-α-induced RhoA transcriptions in human bronchial smooth muscle cells.
Goto K, et al.
Pharmacological Research, 61(5), 466-472 (2010)
Shinya Nagashima et al.
Bioorganic & medicinal chemistry, 15(2), 1044-1055 (2006-10-31)
The STAT6 (signal transducers and activators of transcription 6) protein is activated by interleukin (IL)-4 and IL-13, and plays an important role in T-helper cell 2 (Th2) differentiation. STAT6 might therefore be an excellent therapeutic target for various allergic conditions
Xuechao Jiang et al.
Cell death & disease, 7(12), e2506-e2506 (2016-12-03)
Signal transducer and activator of transcription 6 (STAT6) is involved in epithelial cell growth. However, little is known regarding the STAT6 phosphorylation status in Graves' disease (GD) and its role in thyroid epithelial cells (TECs). In this study, we found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service