Skip to Content
Merck
All Photos(1)

Key Documents

SML1143

Sigma-Aldrich

PTC-209

≥98% (HPLC)

Synonym(s):

N-(2,6-Dibromo-4-methoxyphenyl)-4-(2-methylimidazo[1,2-a]pyrimidin-3-yl)-2-thiazolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13Br2N5OS
CAS Number:
Molecular Weight:
495.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

BrC1=CC(OC)=CC(Br)=C1NC2=NC(C3=C(C)N=C4N3C=CC=N4)=CS2

InChI

1S/C17H13Br2N5OS/c1-9-15(24-5-3-4-20-16(24)21-9)13-8-26-17(22-13)23-14-11(18)6-10(25-2)7-12(14)19/h3-8H,1-2H3,(H,22,23)

InChI key

XVOOCQSWCCRVDY-UHFFFAOYSA-N

Application

PTC-209 has been used as a B lymphoma Mo-MLV insertion region 1 homolog (BMI1) inhibitor to study its effects on:
  • the cell viability of canine osteosarcoma (OSA) cell lines
  • lipidated microtubule-associated protein 1 light chain 3 (LC3B-II) and Sequestosome 1 (SQSTM1) protein levels in ovarian cancer (OvCa) cells
  • cardiac reprogramming efficiency in mouse embryonic fibroblasts (MEF)

Biochem/physiol Actions

PTC-209 inhibits the expression of the oncogene BMI-1 (B lymphoma Mo-MLV insertion region 1 homolog), which encodes a polycomb group RING finger protein involved in cell cycle. PTC-209 lowers the self-renewal properties of colorectal cancer-initiating cells (CICs) The compound inhibits tumor growth, and reduces the levels of CICs present in colon cancer tumor xenografts in mice.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of BMI1 induces autophagy-mediated necroptosis
Dey A, et al.
Autophagy, 12(4), 659-670 (2016)
BMI1 is expressed in canine osteosarcoma and contributes to cell growth and chemotherapy resistance
Shahi M H, et al.
PLoS ONE, 10(6), e0131006-e0131006 (2015)
Mohammad Alzrigat et al.
Oncotarget, 8(61), 103731-103743 (2017-12-22)
Multiple myeloma (MM) is a tumor of plasmablasts/plasma cells (PCs) characterized by the expansion of malignant PCs with complex genetic aberrations in the bone marrow (BM). Recent reports, by us and others, have highlighted the polycomb group (PcG) proteins as
Sami Al-Nadaf et al.
Veterinary and comparative oncology, 20(4), 871-880 (2022-07-15)
The BMI1 proto-oncogene, polycomb ring finger protein (BMI1) is a key component of the epigenetic polycomb repressor complex 1, and has been associated with aggressive behaviour and chemotherapeutic resistance in various malignances including human gliomas. Similar to humans, spontaneous canine
Junan Li et al.
Anticancer research, 40(1), 133-141 (2020-01-02)
Aberrant expression of the BMI1 oncogene has been prevalently found in a variety of human cancers, including cervical cancer. Recent studies have shown that PTC209, a specific BMI1 inhibitor, exhibits high potency in inhibiting the growth of colon, breast, oral

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service