Skip to Content
Merck
All Photos(1)

Key Documents

SML0795

Sigma-Aldrich

PF-670462

≥98% (HPLC)

Synonym(s):

4-[3-Cyclohexyl-5-(4-fluoro-phenyl)-3H-imidazol-4-yl]-pyrimidin-2-ylamine dihydrochloride, PF670462

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20FN5 · 2HCl
CAS Number:
Molecular Weight:
410.32
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)c2nc[n](c2c4nc(ncc4)N)C3CCCCC3.Cl.Cl

InChI

1S/C19H20FN5.2ClH/c20-14-8-6-13(7-9-14)17-18(16-10-11-22-19(21)24-16)25(12-23-17)15-4-2-1-3-5-15;;/h6-12,15H,1-5H2,(H2,21,22,24);2*1H

InChI key

PSNKGVAXBSAHCH-UHFFFAOYSA-N

Biochem/physiol Actions

PF-670462 is a selective inhibitor of the δ- and ε-isoforms of casein kinase I, with IC50 values of 7.7 and 14 nM respectively, and >30 selectivity relative to 42 other kinases tested. Casein kinase Iε phosphorylates PER proteins, which are involved in setting the period of the circadian pacemaker or clock. PF-670462 is potent (IC50 7.7 nM) and effective in vivo (i.e. it induces profound phase delays in circadian periodicity).
PF-670462 is a selective inhibitor of the δ- and ε-isoforms of casein kinase I.
PF-670462 is also termed as 4-[1-cyclohexyl-4-(4-fluorophenyl)-1H-imidazol-5-yl]-2-pyrimidinamine dihydrochloride). It participates in ceramide transfer between Golgi and ER (endoplasmic reticulum) compartments. PF-670462 affects actin filament consolidation. It also hinders the stability at the front edge of cells treated with N-formyl Met-Leu-Phe.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Inhibitor Index: A Desk Reference on Enzyme Inhibitors, Receptor Antagonists, Drugs, Toxins, Poisons, Biologics, and Therapeutic Leads, 1362-1362 (2017)
Combined Pharmacological and Genetic Manipulations Unlock Unprecedented Temporal Elasticity and Reveal Phase-Specific Modulation of the Molecular Circadian Clock of the Mouse Suprachiasmatic Nucleus.
Patton AP, et al.
The Journal of Neuroscience, 36(36), 9326-9341 (2016)
David A Hicks et al.
Neurochemical research, 45(6), 1354-1364 (2019-07-08)
Motor neuron disease (MND) is a progressive neurodegenerative disease with no effective treatment. One of the principal pathological hallmarks is the deposition of TAR DNA binding protein 43 (TDP-43) in cytoplasmic inclusions. TDP-43 aggregation occurs in both familial and sporadic
Lucian B Tomaz et al.
Journal of cell science, 135(21) (2022-10-12)
The gene mutated in colorectal cancer (MCC) encodes a coiled-coil protein implicated, as its name suggests, in the pathogenesis of hereditary human colon cancer. To date, however, the contributions of MCC to intestinal homeostasis and disease remain unclear. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service