Skip to Content
Merck
All Photos(1)

Key Documents

SML0772

Sigma-Aldrich

Curvularin

from Penicillium citrinum, ≥98% (HPLC)

Synonym(s):

2H-3-Benzoxacyclododecin-2,10(1H)-dione, 4,5,6,7,8,9-hexahydro-11,13-dihydroxy-4-methyl-,(4S)-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20O5
CAS Number:
Molecular Weight:
292.33
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

biological source

Penicillium citrinum

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: 1 mg/mL

storage temp.

−20°C

SMILES string

OC1=C(C(CCCCC[C@H](C)O2)=O)C(CC2=O)=CC(O)=C1

InChI

1S/C16H20O5/c1-10-5-3-2-4-6-13(18)16-11(8-15(20)21-10)7-12(17)9-14(16)19/h7,9-10,17,19H,2-6,8H2,1H3/t10-/m0/s1

InChI key

VDUIGYAPSXCJFC-JTQLQIEISA-N

Biochem/physiol Actions

S - Curvularin is a macrocyclic lactone with cytotoxic activity. Curvularin inhibits cell division as well as expression of human inducible nitric oxide synthase (iNOS), an enzyme critically involved in pro-inflammatory immune processes. Curvularin anti-inflammatory activity was demonstrated in chronic inflammatory disease models in mice such as Chronic Induced Arthritis (CIA) as well as in human alveolar epithelial A549/8 cells.
Curvularin is a macrocyclic lactone with cytotoxic activity.

Reconstitution

Soluble in DMSO (1 mg/mL), Chloroform (3 mg/mL) and Dichloromethane (3 mg/mL). Note: Insoluble in water.

Other Notes

Store the product sealed at −20 °C. Under these conditions the product is stable for at least 5 years.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ia I Savchuk et al.
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993), 74(4), 52-56 (2012-10-24)
Silica gel column chromatography (silica gel "L" II kind of activity 100/160 mkm) of the chloroform extract from the cultural filtrate of Penicillium sp. 10-51 gave two fractions (chloroform and chloroform-acetone, 5:1) having biological activity. Recrystallization yielded two compounds. On
A new anthraquinone and cytotoxic curvularins of a Penicillium sp. from the rhizosphere of Fallugia paradoxa of the Sonoran desert.
Jixun Zhan et al.
The Journal of antibiotics, 57(5), 341-344 (2004-08-20)
Qiren Liang et al.
The Journal of organic chemistry, 72(25), 9846-9849 (2007-11-09)
Concise and efficient total syntheses of 11-alpha-methoxycurvularin and 11-beta-methoxycurvularin were accomplished for the first time. The three-component linchpin coupling and intramolecular acylation reactions were key steps, in which we found the spectral data of 11-alpha-methoxycurvularin and 11-beta-methoxycurvularin, reported in the
Nadine Schmidt et al.
Biochemical pharmacology, 79(5), 722-732 (2009-10-27)
Chondrocytes are important for the development and maintenance of articular cartilage. However, both in osteoarthritis (OA) and rheumatoid arthritis (RA) chondrocytes are involved in the process of cartilage degradation and synthesize important immunomodulatory mediators, including nitric oxide (NO) generated by
[Molecular basis of probes for cytoskeletal proteins].
A Kobayashi et al.
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme, 38(11), 1730-1741 (1993-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service