Skip to Content
Merck
All Photos(1)

Key Documents

SML0724

Sigma-Aldrich

Bikaverin

from Fusarium subglutinans, ≥98% (HPLC)

Synonym(s):

6,11-Dihydroxy-3,8-dimethoxy-1-methylbenzo[b]xanthene-7,10, 12-trione, Lycopersin, NSC 215139

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14O8
CAS Number:
Molecular Weight:
382.32
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

Fusarium subglutinans

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: 0.5 mg/mL (may require sonication and heating)
chloroform: 0.5 mg/mL
dichloromethane: 0.5 mg/mL

storage temp.

−20°C

SMILES string

[o]1c2c([c](c4c1cc(cc4C)OC)=O)c(c3c(c2O)C(=O)C(=CC3=O)OC)O

InChI

1S/C20H14O8/c1-7-4-8(26-2)5-10-12(7)17(23)15-18(24)13-9(21)6-11(27-3)16(22)14(13)19(25)20(15)28-10/h4-6,24-25H,1-3H3

InChI key

ZOQMSOSJEWBMHP-UHFFFAOYSA-N

Biochem/physiol Actions

Bikaverin is a red pigment with a polyketide tetracyclic benzoxanthone structure. Bikaverin has an antibiotic activity against some protozoa and fungi and also inhibits Succinate- and NAD-linked respiration in rat mitochondria at 20 mg/mL. At higher concentrations (50 mg/mL) it acts as an oxidative phosphorylation uncoupling agent of tumor cells and isolated rat liver mitochondria. Bikaverin demonstrates antitumor activity on Erlich ascites carcinoma (EAC), leukemia and sarcoma cells.
Bikaverin is a red pigment with a polyketide tetracyclic benzoxanthone structure. Bikaverin has an antibiotic activity against some protozoa and fungi.

Reconstitution

Soluble in DMSO (0.5 mg/mL, may require sonication and heating). For further dilution, water based solutions can be used. Chloroform (0.5 mg/mL) and dichloromethane (0.5 mg/mL).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A G Medentsev et al.
Prikladnaia biokhimiia i mikrobiologiia, 41(5), 573-577 (2005-10-26)
We studied biosynthesis of colored naphthoquinone metabolites by Fusarium decemcellulare, F. graminearum, and F. bulbigenum fungi. F. bulbigenum and F. graminearum synthesized bikaverin and aurofusarin, respectively, which depended on the conditions of cultivation. F. decemcellulare synthesized soluble extracellular naphthoquinones of
Alejandro F Estrada et al.
Fungal genetics and biology : FG & B, 45(5), 705-718 (2008-01-22)
The fungal proteins of the White Collar photoreceptor family, represented by WC-1 from Neurospora crassa, mediate the control by light of different biochemical and developmental processes, such as carotenogenesis or sporulation. Carotenoid biosynthesis is induced by light in the gibberellin-producing
Roberto Rodríguez-Ortiz et al.
Applied microbiology and biotechnology, 85(6), 1991-2000 (2009-10-20)
The fungus Fusarium fujikuroi (Gibberella fujikuroi mating group C) exhibits a rich secondary metabolism that includes the synthesis of compounds of biotechnological interest, such as gibberellins, bikaverin, and carotenoids. The effect of the carbon source on their production was checked
Mayra Mendonça et al.
Food chemistry: X, 12, 100136-100136 (2021-10-19)
Microbial pigments have a distinguished potential for applications in food and pharmaceutical industries, stimulating the research in this field. The present study evaluated the ideal conditions for extracting bikaverin (red pigment) from the biomass of Fusarium oxysporum CCT7620. Among the
Yoon-E Choi et al.
Mycologia, 100(5), 701-709 (2008-10-31)
Fusarium verticillioides (teleomorph Gibberella moniliformis) is an ascomycete known to produce a variety of secondary metabolites, including fumonisins, fusaric acid and bikaverin. These metabolites are synthesized when the fungus is under stress, notably nutrient limitations. To date we have limited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service