Skip to Content
Merck
All Photos(3)

Key Documents

SML0580

Sigma-Aldrich

Nutlin-3a

≥98% (HPLC)

Synonym(s):

(-)-4-(4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydro-1H-imidazole-1-carbonyl)piperazin-2-one, (-)-Nutlin-3, 4-[[(4S,5R)-4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydroimidazol-1-yl]carbonyl]piperazin-2-one, 4-[[(4S,5R)-4,5-Bis(4-chlorophenyl)-4,5-dihydro-2-[4-methoxy-2-(1-methylethoxy)phenyl]-1H-imidazol-1-yl]carbonyl]-2-Piperazinone, Nutlin 3a

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H30Cl2N4O4
CAS Number:
Molecular Weight:
581.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

ligand

nutlin-3a

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -130 to -154° (c=0.5, CDCl3)

reaction suitability

reagent type: ligand

color

white to beige

solubility

DMSO: 5 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(N1CC(NCC1)=O)N2[C@H](C3=CC=C(Cl)C=C3)[C@H](C4=CC=C(Cl)C=C4)N=C2C5=CC=C(OC)C=C5OC(C)C

InChI

1S/C30H30Cl2N4O4/c1-18(2)40-25-16-23(39-3)12-13-24(25)29-34-27(19-4-8-21(31)9-5-19)28(20-6-10-22(32)11-7-20)36(29)30(38)35-15-14-33-26(37)17-35/h4-13,16,18,27-28H,14-15,17H2,1-3H3,(H,33,37)/t27-,28+/m0/s1

InChI key

BDUHCSBCVGXTJM-WUFINQPMSA-N

Application

Nutlin-3a has been used as a tumor suppressor p53(TP53) stabilizer and as an inhibitor of the mouse double minute 2 homolog (MDM2)–p53 interaction.

Biochem/physiol Actions

Nutlin-3a [(-)-Nutlin] is a more potent diastereoisomer of racemic MDM2 antagonist Nutlin-3. It was called enantiomer-a since it elutes as the first peak from chiral purification of racemic nutlin-3. Nutlin-3a is a potent inhibitor of MDM2 (mouse double minute 2) binding to p53 that induces the expression of p53 regulated genes, and shows potent antiproliferative activity in cells expressing functional p53. Nutlin-3a is 150 times more potent than Nutlin-3b.
Nutlin-3a [(-)-Nutlin] is a potent inhibitor of MDM2 (mouse double minute 2) binding to p53; Mdm2 antagonist.
Nutlin-3a is a potent nongenotoxic drug. It acts as a tumor suppressor p53 (TP53) activator and stabilizer.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conservation and Divergence of p53 Oscillation Dynamics across Species.
Jacob S O, et al.
Cell Systems, 5(4), 410-417 (2017)
Manasi Das et al.
Nanomedicine : nanotechnology, biology, and medicine, 11(2), 379-389 (2014-09-23)
The promising proposition of multifunctional nanoparticles for cancer diagnostics and therapeutics has inspired the development of theranostic approach for improved cancer therapy. Moreover, active targeting of drug carrier to specific target site is crucial for providing efficient delivery of therapeutics
Long non-coding RNA NEAT1-modulated abnormal lipolysis via ATGL drives hepatocellular carcinoma proliferation.
Liu X, et al.
Molecular Cancer, 17(1), 90-90 (2018)
Emma Haapaniemi et al.
Nature medicine, 24(7), 927-930 (2018-06-13)
Here, we report that genome editing by CRISPR-Cas9 induces a p53-mediated DNA damage response and cell cycle arrest in immortalized human retinal pigment epithelial cells, leading to a selection against cells with a functional p53 pathway. Inhibition of p53 prevents
Gozde Korkmaz et al.
Nature biotechnology, 34(2), 192-198 (2016-01-12)
Systematic identification of noncoding regulatory elements has, to date, mainly relied on large-scale reporter assays that do not reproduce endogenous conditions. We present two distinct CRISPR-Cas9 genetic screens to identify and characterize functional enhancers in their native context. Our strategy

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service