Skip to Content
Merck
All Photos(1)

Key Documents

SML0568

Sigma-Aldrich

3-AP

≥98% (HPLC)

Synonym(s):

3-AP, 3-Aminopyridine-2-carboxaldehyde thiosemicarbazone, PAN-811

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N5S
CAS Number:
Molecular Weight:
195.24
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 meq/mL, clear

storage temp.

2-8°C

SMILES string

S=C(N\N=C\c1ncccc1N)N

InChI

1S/C7H9N5S/c8-5-2-1-3-10-6(5)4-11-12-7(9)13/h1-4H,8H2,(H3,9,12,13)/b11-4+

InChI key

XMYKNCNAZKMVQN-NYYWCZLTSA-N

Biochem/physiol Actions

3-Aminopyridine-2-carboxaldehyde thiosemicarbazone (3-AP) is a ribonucleotide reductase inhibitor and iron chelator with anti-tumor activity.
3-Aminopyridine-2-carboxaldehyde thiosemicarbazone (3-AP) is a ribonucleotide reductase inhibitor and iron chelator with anti-tumor activity.
3-aminopyridine carboxaldehyde thiosemicarbazone (3-AP) has a IC50 value of 0.3μM. It exhibits anti-proliferative activity in preclinical models of cancer, such as lung cancer. It also has an ability to increase the cytotoxicity, intracellular uptake and DNA incorporation of gemcitabine in vitro.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R A Finch et al.
Advances in enzyme regulation, 39, 3-12 (1999-09-02)
The enzyme RR catalyzes the conversion of ribonucleoside diphosphates to their deoxyribonucleotide counterparts. RR is critical for the generation of the cytosine, adenine, and guanine deoxyribonucleotide 5'-triphosphate building blocks of DNA, which are present in cells as exceedingly small intracellular
Jia Shou et al.
Molecular cell, 71(4), 498-509 (2018-07-24)
Chromosomal rearrangements including large DNA-fragment inversions, deletions, and duplications by Cas9 with paired sgRNAs are important to investigate genome structural variations and developmental gene regulation, but little is known about the underlying mechanisms. Here, we report that disrupting CtIP or
Éva A Enyedy et al.
Dalton transactions (Cambridge, England : 2003), 40(22), 5895-5905 (2011-04-28)
Stoichiometry and stability of Ga(III), Fe(III), Fe(II) complexes of Triapine and five related α-N heterocyclic thiosemicarbazones with potential antitumor activity have been determined by pH-potentiometry, UV-vis spectrophotometry, (1)H NMR spectroscopy, and spectrofluorimetry in aqueous solution (with 30% DMSO), together with
Charles A Kunos et al.
Radiation research, 176(4), 425-433 (2011-07-16)
Cells generate 2'-deoxyribonucleoside triphosphates (dNTPs) for both replication and repair of damaged DNA predominantly through de novo reduction of intracellular ribonucleotides by ribonucleotide reductase (RNR). Cells can also salvage deoxynucleosides by deoxycytidine kinase/thymidine kinase 1 in the cytosol or by
J Li et al.
Current medicinal chemistry, 8(2), 121-133 (2001-02-15)
The reductive conversion of ribonucleotides to deoxyribonucleotides by ribonucleotide reductase (RR) is a crucial and rate-controlling step in the pathway leading to the biosynthesis of DNA, since deoxyribonucleotides are present in extremely low levels in mammalian cells. Mammalian ribonucleotide reductase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service