Skip to Content
Merck
All Photos(1)

Key Documents

SML0543

Sigma-Aldrich

p38 MAP Kinase Inhibitor IV

≥98% (HPLC)

Synonym(s):

2,2′-Sulfonyl-bis-(3,4,6-trichlorophenol), B10, MT4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4Cl6O4S
CAS Number:
Molecular Weight:
456.94
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(c2c(c(cc(c2O)Cl)Cl)Cl)c1c(c(cc(c1O)Cl)Cl)Cl

InChI

1S/C12H4Cl6O4S/c13-3-1-5(15)9(19)11(7(3)17)23(21,22)12-8(18)4(14)2-6(16)10(12)20/h1-2,19-20H

InChI key

AKUKHICVNCCQHN-UHFFFAOYSA-N

Application

p38 MAP Kinase Inhibitor IV has been used for blocking p38α kinase activation in order to examine if HOXB7 is involved in the regulation of migration and proliferation process via AKT/MAPK signaling.

Biochem/physiol Actions

p38 MAP Kinase Inhibitor IV is an ATP-competitive inhibitor of p38α/β MAPK with IC50 values of 130 nM for p38α and 550 nM for p38β. It is much less active with ≤23% inhibition at 1 μM against p38γ/σ, ERK1/2, and JNK1/2/3. Shown to be more effective than SB 203580 in inhibiting LPS-induced IL-1β release from hPBMC (100% vs. 50% inhibition with 100 μM inhibitor). A recent study showed that p38 MAP Kinase Inhibitor IV could consistently and significantly enhance reprogramming and iPS cell generation from somatic cells.
p38 MAP Kinase Inhibitor IV is an ATP-competitive inhibitor of p38α/β MAPK.

Features and Benefits

This compound is featured on the MAPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xujun He et al.
Oncotarget, 8(1), 1247-1261 (2016-12-03)
Increased expression of HOXB7 has been reported to correlate with the progression in many cancers. However, the specific mechanism by which it promotes the evolution of gastric cancer (GC) is poorly understood.In this study, we sought to investigate the role
Jong-Hwan Woo et al.
Particle and fibre toxicology, 20(1), 2-2 (2023-01-10)
Polypropylene (PP) is used in various products such as disposable containers, spoons, and automobile parts. The disposable masks used for COVID-19 prevention mainly comprise PP, and the disposal of such masks is concerning because of the potential environmental pollution. Recent
Denis Nchang Che et al.
International immunopharmacology, 83, 106403-106403 (2020-03-21)
IL-31 and IL-33 are cytokines, which are expressed in many inflammatory and pathological disorders, thus suggesting an IL-31/IL-33 axis interaction in pathological diseases. Luteolin from natural products is known for its anti-inflammatory activities associated with the regulation of inflammatory signaling
Hanna Niehues et al.
JID innovations : skin science from molecules to population health, 2(1), 100066-100066 (2022-02-12)
Psoriasis and atopic dermatitis are chronic inflammatory skin diseases characterized by keratinocyte (KC) hyperproliferation and epidermal acanthosis (hyperplasia). The milieu of disease-associated cytokines and soluble factors is considered a mitogenic factor; however, pinpointing the exact mitogens in this complex microenvironment
Douglas P Jasmer et al.
Pharmaceuticals (Basel, Switzerland), 14(7) (2021-07-03)
Establishing methods to investigate treatments that induce cell death in parasitic nematodes will promote experimental approaches to elucidate mechanisms and to identify prospective anthelmintics capable of inducing this outcome. Here, we extended recent progress on a method to monitor cell

Articles

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service