Skip to Content
Merck
All Photos(1)

Documents

SML0446

Sigma-Aldrich

A-419259 trihydrochloride

≥98% (HPLC)

Synonym(s):

7-[Trans-4-(4-methyl-1-piperazinyl)cyclohexyl]-5-(4-phenoxyphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine trihydrochloride, A419259 trihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H34N6O · 3HCl
CAS Number:
Molecular Weight:
592.00
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 10 mg/mL (clear solution)

storage temp.

room temp

SMILES string

Cl.Cl.Cl.CN1CCN(CC1)[C@H]2CC[C@@H](CC2)n3cc(-c4ccc(Oc5ccccc5)cc4)c6c(N)ncnc36

InChI

1S/C29H34N6O.3ClH/c1-33-15-17-34(18-16-33)22-9-11-23(12-10-22)35-19-26(27-28(30)31-20-32-29(27)35)21-7-13-25(14-8-21)36-24-5-3-2-4-6-24;;;/h2-8,13-14,19-20,22-23H,9-12,15-18H2,1H3,(H2,30,31,32);3*1H/t22-,23-;;;

InChI key

ALRMEQIQFCUAMR-KOONDFAVSA-N

Biochem/physiol Actions

A-419259 is a potent inhibitor of Src family kinases. IC50 values for src, lck, blk, csk, fyn and lyn range between 15 and 50 nM.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Src is the original member of a group of non-receptor protein-tyrosine kinases termed the Src family kinases (SFKs). Learn about the SFKs that, in humans, consist of eight members, a small group of atypical members, and two related kinases.

Src is the original member of a group of non-receptor protein-tyrosine kinases termed the Src family kinases (SFKs). Learn about the SFKs that, in humans, consist of eight members, a small group of atypical members, and two related kinases.

Src is the original member of a group of non-receptor protein-tyrosine kinases termed the Src family kinases (SFKs). Learn about the SFKs that, in humans, consist of eight members, a small group of atypical members, and two related kinases.

Src is the original member of a group of non-receptor protein-tyrosine kinases termed the Src family kinases (SFKs). Learn about the SFKs that, in humans, consist of eight members, a small group of atypical members, and two related kinases.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service