Skip to Content
Merck
All Photos(1)

Key Documents

SML0187

Sigma-Aldrich

CJ-13610

≥98% (HPLC)

Synonym(s):

Tetrahydro-4-[3-[[4-(2-methyl-1H-imidazol-1-yl)phenyl]thio]phenyl]-2H-pyran-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23N3O2S
CAS Number:
Molecular Weight:
393.50
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

Cc1nccn1-c2ccc(Sc3cccc(c3)C4(CCOCC4)C(N)=O)cc2

InChI

1S/C22H23N3O2S/c1-16-24-11-12-25(16)18-5-7-19(8-6-18)28-20-4-2-3-17(15-20)22(21(23)26)9-13-27-14-10-22/h2-8,11-12,15H,9-10,13-14H2,1H3,(H2,23,26)

InChI key

VPTONMHDLLMOOV-UHFFFAOYSA-N

Application

CJ-13610 may be used to study 5-lipoxygenase-mediated cell signaling.

Biochem/physiol Actions

CJ-13,610 is a potent inhbititor of 5-lipoxygenase (5-LO) activity (IC50 = 70 nM).
CJ-13610 inhibits the biosynthesis of leukotriene B4 and regulates the IL-6 mRNA expression in macrophages. It reduces fibrosis and necroinflammation of liver in carbon tetrachloride-treated mice.

Features and Benefits

This compound is featured on the Leukotriene Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service