Skip to Content
Merck
All Photos(1)

Documents

SML0087

Sigma-Aldrich

MEGX

≥95% (HPLC)

Synonym(s):

MGEX, Monoethylglycinexylidide, N-(2,6-dimethylphenyl)-2-(ethylamino)acetamide, Norlidocaine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N2O
CAS Number:
Molecular Weight:
206.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(NC(CNCC)=O)C(C)=CC=C1

InChI

1S/C12H18N2O/c1-4-13-8-11(15)14-12-9(2)6-5-7-10(12)3/h5-7,13H,4,8H2,1-3H3,(H,14,15)

InChI key

WRMRXPASUROZGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

MEGX has been used in an approach to improve the yield of N-dealkylated lidocaine.

Biochem/physiol Actions

MEGX is generated by hepatic cytochrome P-450 system upon oxidative de-ethylation. Measurement of MEGX by immunoassay contributes to liver function test. MEGX possesses antiarrhythmic action and therefore extends lidocaine effect.
Monoethylglycinexylidide (MEGX) is an active metabolite of lidocaine.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Venkatachalam et al.
New Zealand veterinary journal, 67(5), 228-233 (2019-04-30)
To investigate the analgesic efficacy of articaine hydrochloride for antler removal in red deer (Cervus elaphus) following S/C administration as a ring block, and to quantify the residue concentrations of articaine compared to lignocaine in the harvested antlers. Articaine hydrochloride
Georgette Oni et al.
Aesthetic surgery journal, 30(6), 853-858 (2010-12-07)
Topical lidocaine is a common form of anesthesia for a wealth of procedures across a large number of disciplines, including laser treatments. Preparations can be purchased over the counter with no prescription necessary. It is considered a safer and more
Hamdah M Al Nebaihi et al.
Journal of pharmaceutical sciences, 109(2), 1199-1210 (2019-11-17)
The effects of a high-fat diet on mRNA and protein of cytochrome P450 (CYP) enzymes in rats and mice and its impact on lidocaine deethylation to its main active metabolite, monoethylglycinexylidide (MEGX), in rats were investigated. The effect of a
Robert Werdehausen et al.
Anesthesiology, 116(1), 147-158 (2011-12-03)
Lidocaine exerts antinociceptive effects when applied systemically. The mechanisms are not fully understood but glycinergic mechanisms might be involved. The synaptic glycine concentration is controlled by glycine transporters. Whereas neurons express two types of glycine transporters, astrocytes specifically express glycine
Dinakaran Venkatachalam et al.
Animals : an open access journal from MDPI, 8(8) (2018-08-22)
This study determined the convulsant plasma concentrations and pharmacokinetic parameters following cornual nerve block and compared the results to recommend a safe dose of lidocaine hydrochloride for goat kids. The plasma concentrations of lidocaine and monoethylglycinexylidide (MGX) were quantified using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service