Skip to Content
Merck
All Photos(1)

Documents

SMB00112

Sigma-Aldrich

Andrographiside

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H40O10
CAS Number:
Molecular Weight:
512.59
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C[C@@]12CC[C@@H](O)[C@@](C)(CO[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@H]1CCC(=C)[C@H]2C\C=C4/C[C@@H](O)OC4=O

InChI

1S/C26H40O10/c1-13-4-7-17-25(2,15(13)6-5-14-10-19(29)36-23(14)33)9-8-18(28)26(17,3)12-34-24-22(32)21(31)20(30)16(11-27)35-24/h5,15-22,24,27-32H,1,4,6-12H2,2-3H3/b14-5+/t15-,16-,17+,18-,19+,20-,21+,22-,24-,25+,26+/m1/s1

InChI key

ODNIGBWMYZIVBO-AZWZBXPUSA-N

General description

Andrographiside is a labdane diterpenoid glucoside found in Andrographis paniculata, which is an annual herbaceous plant traditionally used to treat infections in Southern Asia.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Li et al.
Cellular & molecular immunology, 6(5), 381-385 (2009-11-06)
Andrographolide from traditional Chinese herbal medicines previously showed it possesses a strong anti-inflammatory activity. In present study, we investigated whether Andrographolide could inhibit allergen-induced airway inflammation and airways hyper-responsiveness and explored the mechanism of Andrographolide on allergen-induced airway inflammation and
[Isolation and structure of two new diterpenoid glucosides from Andrographis paniculata Nees].
C Q Hu et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 17(6), 435-440 (1982-06-01)
[Pharmacological studies on thirteen kinds of injections from Andrographis paniculata. I. Antipyretic, anti-inflammatory effects and toxicity].
W L Deng
Zhong yao tong bao (Beijing, China : 1981), 10(7), 38-42 (1985-07-01)
[Comprehensive evaluation of cuanhuning injection].
Y Huang
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 26(6), 427-429 (2003-01-17)
J Melchior et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 7(5), 341-350 (2000-11-18)
Two randomized double-blind, placebo-controlled parallel group clinical trials were performed to investigate the effect of a standardized extract (SHA-10) of Andrographis paniculata fixed combination (Kan jang) in the treatment of uncomplicated upper-respiratory tract infections. 46 patients in the pilot study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service