Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700637

Sigma-Aldrich

Monoclonal Anti-HLA Class I, low endotoxin antibody produced in mouse

clone W6/32, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

W6/32, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

impurities

≤1 EU/μg endotoxin (LAL test)

isotype

IgG2a

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

Human leukocyte antigen A classical class I family consists of three members including HLA-A, HLA-B and HLA-C. Members of classical class I major histocompatibility complex (MHC) contain a peptide bound to a transmembrane glycoprotein subunit, the heavy chain, in association with a soluble light chain called β2-microglobulin. Class I MHC molecules are present in changeable volumes on all nucleated cells.
The antibody W6/32 recognises MHC Class I molecules (MHC Class Ia) that are expressed on the surface of all human nucleated cell types. The antibody W6/32 is a valuable reagent for analysing variations in HLA class I expression in different disease states e.g. liver disease, muscular dystrophy, inflammatory myopathy and other neuromuscular disorders.

The previously assigned protein identifier P01891 has been merged into P04439. Full details can be found on the UniProt database.

Immunogen

Membrane of human tonsil cells

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 2-8 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

Human leukocyte antigen A class I downregulation is implicated in tumor invasion and development. Genetic variations in the class I major histocompatibility complex (MHC) is associated with the development of melanoma. HLA-viral peptide interaction plays a major role in the regulation of human immunodeficiency virus (HIV) infection.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in azide free phosphate buffered saline, pH 7.4; 0.2 um filter sterilized. Endotoxin level is less than 0.01 EU/ug of the protein, as determined by the LAL test.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genes regulating HLA class I antigen expression in T-B lymphoblast hybrids
Salter RD
Immunogenetics, 21, 235-246 (1985)
Marco Bolis et al.
Cancers, 12(5) (2020-05-10)
All-trans retinoic acid (ATRA), a recognized differentiating agent, has significant potential in the personalized/stratified treatment of breast cancer. The present study reports on the molecular mechanisms underlying the anti-tumor activity of ATRA in breast cancer. The work is based on
Loss of HLA class I antigens by melanoma cells: molecular mechanisms, functional significance and clinial relevance
Ferrone S and Marincola FM
Immunology Today, 16, 487-494 (1995)
Paulina Moreno-Layseca et al.
Nature cell biology, 23(10), 1073-1084 (2021-10-08)
Spatially controlled, cargo-specific endocytosis is essential for development, tissue homeostasis and cancer invasion. Unlike cargo-specific clathrin-mediated endocytosis, the clathrin- and dynamin-independent endocytic pathway (CLIC-GEEC, CG pathway) is considered a bulk internalization route for the fluid phase, glycosylated membrane proteins and
Implications for immunosurveillance of altered HLA class I phenotypes in human tumours.
Garrido F
Immunology Today, 18, 89-95 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service