Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700632

Sigma-Aldrich

Monoclonal Anti-HLA Class I antibody produced in mouse

clone MEM-81, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-81, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

Human leukocyte antigen A classical class I family consists of three members including HLA-A, HLA-B and HLA-C. Members of classical class I major histocompatibility complex (MHC) contain a peptide bound to a transmembrane glycoprotein subunit, the heavy chain, in association with a soluble light chain called β2-microglobulin. Class I MHC are present in changeable volumes on all nucleated cells.
The antibody MEM-81 reacts with human classical MHC Class I molecules in native cell-surface forms. MHC Class I molecules (MHC Class Ia) are expressed on the surface of human nucleated cell types.

The previously assigned protein identifier P01891 has been merged into P04439. Full details can be found on the UniProt database.

Immunogen

Leukocytes of a patient suffering with LGL type leukemia

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 1 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

Human leukocyte antigen A class I downregulation is implicated in tumor invasion and development. Genetic variations in the class I major histocompatibility complex (MHC) is associated with the development of melanoma. HLA-viral peptide interaction plays a major role in the regulation of human immunodeficiency virus (HIV) infection.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genes regulating HLA class I antigen expression in T-B lymphoblast hybrids
Salter RD
Immunogenetics, 21, 235-246 (1985)
Loss of HLA class I antigens by melanoma cells: molecular mechanisms, functional significance and clinial relevance
Ferrone S and Marincola FM
Immunology Today, 16, 487-494 (1995)
The Major Genetic Determinants of HIV-1 Control Affect HLA Class I Peptide Presentation
Pereyra F
Science, 330, 1551-1557 (2010)
Implications for immunosurveillance of altered HLA class I phenotypes in human tumours.
Garrido F
Immunology Today, 18, 89-95 (1997)
Recognition of human histocompatibility leukocyte antigen (HLA)-E complexed with HLA class I signal sequence-derived peptides by CD94/NKG2 confers protection from natural killer cell-mediated lysis.
Borrego F
The Journal of Experimental Medicine, 187, 813-818 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service