Skip to Content
Merck
All Photos(2)

Key Documents

SAB4504326

Sigma-Aldrich

Anti-phospho-Vitamin D Receptor (pSer208) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer208)

Gene Information

human ... VDR(7421)

General description

VDR (vitamin D receptor) or vitamin D3 receptor is a member of the steroid and thyroid hormone receptors superfamily of trans-acting transcriptional regulatory factors. It is an intracellular polypeptide of 50-60 kDa. This receptor is composed of a highly conserved cysteine, lysine, and arginine-rich DNA-binding domain, and a hydrophobic ligand-binding domain in its C-terminal. It shares size and sequence similarity to thyroid hormone receptor. VDR gene is localized to human chromosome 12q12-q14.

Immunogen

The antiserum was produced against synthesized peptide derived from human Vitamin D Receptor around the phosphorylation site of Ser208.

Immunogen Range: 181-230

Biochem/physiol Actions

VDR (vitamin D receptor) binds with its ligand vitamin D3, and acts upon the target cell nuclei to induce various biological effects. Vitamin D3 is also thought to have immunomodulatory effects, and gene polymorphisms are linked with pertussis. The variants might affect the immune response and clinical outcome of Bordetella pertussis infection.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of vitamin D receptor polymorphism with susceptibility to symptomatic pertussis
Han WGH, et al.
PLoS ONE, 11(2), e0149576-e0149576 (2016)
Association between VDR FokI polymorphism and intervertebral disk degeneration.
Zhao J, et al.
Genomics, Proteomics & Bioinformatics, 13(6), 371-376 (2015)
Cloning and expression of full-length cDNA encoding human vitamin D receptor.
Baker A R, et al.
Proceedings of the National Academy of Sciences of the USA, 85(10), 3294-3298 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service