Skip to Content
Merck
All Photos(2)

Key Documents

SAB4504246

Sigma-Aldrich

Anti-phospho-IRAK1 (pThr209) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-IRAK, Anti-pelle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

76 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr209)

Gene Information

human ... IRAK1(3654)

General description

Interleukin 1 receptor associated kinase 1 (IRAK1) is a serine/threonine kinase encoded by the gene with 14 exons mapped to human chromosome xq28. The encoded protein belongs to the IRAK family. IRAK1is characterized with a death domain at its N-terminal end which is involved in protein interaction.

Immunogen

The antiserum was produced against synthesized peptide derived from human IRAK1 around the phosphorylation site of Thr209.

Immunogen Range: 175-224

Application

Anti-phospho-IRAK1 (pThr209) antibody produced in rabbit has been used in immunoprecipitation assay.

Biochem/physiol Actions

Interleukin 1 receptor associated kinase 1 (IRAK1) is a vital element of the interleukin-1 receptor (IL-1R) signaling pathway. IRAK1 functions as a downstream effector molecule of the toll like receptor (TLR) signaling pathway, implicated in inflammation, autoimmunity and cancer. IRAK1 is associated with systemic lupus erythematosus (SLE) and it is overexpressed in various cancer types including lung cancer. IRAK1, is considered to be a potential therapeutic target in myelodysplastic syndrome (MDS), acute myeloid leukemia (AML), triple-negative breast cancer (TNBC) metastasis and paclitaxel resistance. Increased expression of IRAK1 leads to head and neck squamous cell carcinomas (HNSCCs). Therefore, inhibition of IRAK1 might be a new therapeutic strategy in HNSCC.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiuling Zhang et al.
International journal of clinical and experimental pathology, 7(11), 8096-8104 (2015-01-01)
The interleukin-1 receptor associated kinases 1 (IRAK1) is a down stream effector molecule of the toll like receptor (TLR) signaling pathway, which is involved in inflammation, autoimmunity and cancer. However, the role of IRAK1 in lung cancer remains unclarified. Herein
Allie K Adams et al.
Oncotarget, 6(41), 43395-43407 (2015-11-04)
The chromatin-binding DEK protein was recently reported to promote the growth of HPV+ and HPV- head and neck squamous cell carcinomas (HNSCCs). Relevant cellular and molecular mechanism(s) controlled by DEK in HNSCC remain poorly understood. While DEK is known to
Yosuke Tanaka et al.
Nature communications, 13(1), 271-271 (2022-01-14)
Leukemia stem cells (LSCs) in chronic myeloid leukemia (CML) are quiescent, insensitive to BCR-ABL1 tyrosine kinase inhibitors (TKIs) and responsible for CML relapse. Therefore, eradicating quiescent CML LSCs is a major goal in CML therapy. Here, using a G0 marker
Devram S Ghorpade et al.
Nature, 555(7698), 673-677 (2018-03-22)
Obesity-induced metabolic disease involves functional integration among several organs via circulating factors, but little is known about crosstalk between liver and visceral adipose tissue (VAT). In obesity, VAT becomes populated with inflammatory adipose tissue macrophages (ATMs). In obese humans, there
Baohuan Cai et al.
Molecular cancer, 20(1), 165-165 (2021-12-16)
Stem Cell leukemia/lymphoma syndrome (SCLL) presents as a myeloproliferative disease which can progress to acute myeloid leukemia and is associated with the coincident development of B-cell and T-cell lymphomas. SCLL is driven by the constitutive activation of fibroblast growth factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service