Skip to Content
Merck
All Photos(3)

Key Documents

SAB4501746

Sigma-Aldrich

Anti-Galectin 3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

35 kDa lectin, CBP 35, Carbohydrate binding protein 35, GALBP, Galactose-specific lectin 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 26 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LGALS3(3958)

Related Categories

General description

Anti-Galectin 3 Antibody detects endogenous levels of total Galectin 3 protein.
Galectin 3 is a carbohydrate-binding protein. It is abundantly expressed in the extracellular matrix, the cytoplasm and the nucleus. It contains an intrinsically disordered N-terminal domain and a canonical CRD (carbohydrate recognition domain). It is mapped to human chromosome 14q22.

Immunogen

The antiserum was produced against synthesized peptide derived from human Galectin 3.

Immunogen Range: 141-190

Biochem/physiol Actions

Galectin 3 controls cell activation, migration, adhesion and signaling. It plays a role in cancer progression after it is initiated. Expression of galectin-3 regulates apoptosis and development of T-cell. It participates in several physiological and pathological processes like pre-mRNA splicing, neoplastic transformation and immune responses.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping-Ping Lang et al.
Laboratory investigation; a journal of technical methods and pathology, 100(3), 378-386 (2019-09-19)
Monocyte and adhesion infiltration into the arterial subendothelium are initial steps in hypertension development. The endothelial intercellular adhesion molecule-1 (ICAM-1) has been implicated in the recruitment and adhesion of leukocytes in several cardiac diseases. However, the role of ICAM-1 in
The intrinsically disordered N-terminal domain of galectin-3 dynamically mediates multisite self-association of the protein through fuzzy interactions.
Lin YH, et al.
The Journal of Biological Chemistry, 292(43), 17845-17856 (2017)
Expression of galectin-3 modulates T-cell growth and apoptosis.
Yang RY, et al.
Proceedings of the National Academy of Sciences of the USA, 93(13), 6737-6742 (1996)
LGALS3 (lectin, galactoside-binding, soluble, 3).
Nangia-Makker P, et al.
Atlas of Genetics and Cytogenetics in Oncology and Haematology, 15(6) (2011)
József Murányi et al.
International journal of molecular sciences, 20(22) (2019-11-14)
Several promising anti-cancer drug-GnRH (gonadotropin-releasing hormone) conjugates have been developed in the last two decades, although none of them have been approved for clinical use yet. Crizotinib is an effective multi-target kinase inhibitor, approved against anaplastic lymphoma kinase (ALK)- or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service