Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501645

Sigma-Aldrich

Anti-VEGFR2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

FLK1, KDR, kinase insert domain receptor, protein-tyrosine kinase receptor Flk-1, vascular endothelial growth factor receptor 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 151 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KDR(3791)

Related Categories

General description

Anti-VEGFR2 Antibody detects endogenous levels of total VEGFR2 protein.
Vascular endothelial growth factor receptor 2 (VEGFR2), also known as kinase insert domain-containing receptor (KDR), is encoded by the gene mapped to human chromosome 4q12. The encoded protein belongs to the VEGF receptor family.

Immunogen

The antiserum was produced against synthesized peptide derived from human VEGFR2.

Immunogen Range: 917-966

Application

Anti-VEGFR2 antibody produced in rabbit has been used in histochemical analysis of hydrogels injected subcutaneously.

Biochem/physiol Actions

Vascular endothelial growth factor receptor 2 (VEGFR2) and its ligand VEGF play a vital role in angiogenesis and inflammation. It also facilitates autocrine stimulation of VEGF. VEGFR2 is implicated in progression of various types of cancers such as gliomas, malignant peripheral nerve sheath tumors, triple-negative breast cancer (TNBC) and gastrointestinal stromal tumors (GISTs).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Whitney L Stoppel et al.
Journal of biomedical materials research. Part A, 104(12), 3058-3072 (2016-10-28)
Heart failure is the leading cause of death in the United States and rapidly becoming the leading cause of death worldwide. While pharmacological treatments can reduce progression to heart failure following myocardial infarction, there still exists a need for new
Elastic, silk-cardiac extracellular matrix hydrogels exhibit time-dependent stiffening that modulates cardiac fibroblast response
Stoppel WL, et al.
Journal of Biomedical Materials Research Part A, 104(12), 3058-3072 (2016)
The Importance of VEGF-KDR Signaling Pathway Genes should Not Be Ignored When the Risk of Developing Multiple Sclerosis is Taken into Consideration.
Mazdeh M, et al.
Journal of Molecular Neuroscience, 62(1), 73-78 (2017)
Phospholipase C gamma 1 (PLCG1) R707Q mutation is counterselected under targeted therapy in a patient with hepatic angiosarcoma
Prenen H, et al.
Oncotarget, 6(34), 36418-36418 (2015)
Molecular biology of the VEGF and the VEGF receptor family.
Clauss M
Seminars in Thrombosis and Hemostasis, 26, 561-570 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service