Skip to Content
Merck
All Photos(2)

Documents

SAB4501295

Sigma-Aldrich

Anti-GLUR2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

GluR-2, GluR-B, GluR-K2, Glutamate receptor 2, Glutamate receptor ionotropic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 98 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRIA2(2891)

General description

Anti-GLUR2 Antibody detects endogenous levels of total GLUR2 protein.
GLUR2 (glutamate receptor 2) is coded by gria2 gene. It is mapped to human chromosome 4q32. It is abundantly expressed in the brain.

Immunogen

The antiserum was produced against synthesized peptide derived from human GluR2.

Immunogen Range: 834-883

Application

Anti-GLUR2, C-Terminal antibody has been used in electron microscopy.

Biochem/physiol Actions

Presence of the glutamate receptor 2 (GluR2) subunit prevents calcium influx through AMPA-receptor complexes. Deletion of this subunit results in enhanced hippocampal long-term potentiation. As such, AMPA receptors play crucial roles in normal neuronal activity, including synaptic plasticity, synaptic scaling, homeostatic synaptic plasticity, learning and memory, as well as in synaptogenesis and the formation of neuronal circuitry.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Involvement of AMPA receptor GluR2 subunits in stimulus-reward learning: evidence from glutamate receptor gria2 knock-out mice
Mead AN and Stephens DN
The Journal of Neuroscience, 23 (2003)
Excitatory synaptic function and plasticity is persistently altered in ventral tegmental area dopamine neurons after prenatal ethanol exposure
Hausknecht K, et al.
Neuropsychopharmacology, 40 (2015)
A nomenclature for ligand-gated ion channels
Collingridge GL, et al.
Neuropharmacology, 56 (2009)
Hu Peng et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 44(38) (2024-08-16)
Ribbon synapses of inner hair cells (IHCs) are uniquely designed for ultrafast and indefatigable neurotransmission of the sound. The molecular machinery ensuring the efficient, compensatory recycling of the synaptic vesicles (SVs), however, remains elusive. This study showed that hair cell
Specificity protein 4 (Sp4) regulates the transcription of AMPA receptor subunit GluA2 (Gria2)
Priya A, et al.
Biochimica et Biophysica Acta (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service