Skip to Content
Merck
All Photos(3)

Documents

SAB4500490

Sigma-Aldrich

Anti-p38 MAPK antibody produced in rabbit

affinity isolated antibody

Synonym(s):

MAP kinase MXI2, MAP kinase p38, MAPK14, kinase p38-α

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 41 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPK14(1432)

General description

Anti-p38 MAPK Antibody detects endogenous levels of total p38 MAPK protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human p38 MAPK.

Immunogen Range: 288-337

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miaoqing Ye et al.
Environmental toxicology, 39(2), 551-561 (2023-07-12)
The mechanisms underlying the progression of non-alcoholic fatty liver disease (NAFLD) into hepatocellular carcinoma (HCC) remains confusing and the therapeutics approaches are also challenging. Here, we aimed to investigate the effects of scoparone on the treatment of HCC stemmed from
Alexey I Goncharov et al.
Biochemistry. Biokhimiia, 86(11), 1446-1460 (2021-12-16)
Progesterone and its synthetic analogues act on cells through different types of receptors, affecting proliferation and apoptosis. These compounds exert their effect through the nuclear receptors and the insufficiently studied membrane progesterone receptors (mPRs) belonging to the progestin and adiponectin
Zhiyong Zhang et al.
Environmental toxicology, 36(8), 1557-1566 (2021-05-07)
Delphinidin is an anthocyanin that belongs to the group of flavonoids that exert numerous biological activities. However, the molecular mechanisms underlying the anticancer effects of delphinidin remain poorly understood. In our study we analyzed delphinidin modulate STAT-3 and MAPKinase signaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service