Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300700

Sigma-Aldrich

Anti-FASN antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FAS, Anti-Fatty Acid Synthase, Anti-OA-519

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~273 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(E-P-R-V-S)

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FASN(2194)

Related Categories

General description

The antibody detects endogenous level of total Fatty Acid Synthase (FASN) protein.

Immunogen

Peptide sequence around aa. 2503-2507 (E-P-R-V-S), according to the protein NP_004095.4

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Fatty acid synthetase catalyzes the formation of long-chain fatty acids from acetyl-CoA, malonyl-CoA and NADPH. This multifunctional protein has 7 catalytic activities and an acyl carrier protein.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojia Chen et al.
Heliyon, 9(12), e22785-e22785 (2023-12-13)
Methyl protodioscin (MPD) is the main component of total diosgenin, which was reported to reduce cholesterol and triglyceride levels potentially. This study aimed to investigate the beneficial effects of MPD against lipid disorder in hyperlipidemic gerbils induced by a high-fat
Krithika P Karthigeyan et al.
The Journal of biological chemistry, 297(5), 101272-101272 (2021-10-05)
Mammalian cells acquire fatty acids (FAs) from dietary sources or via de novo palmitate production by fatty acid synthase (FASN). Although most cells express FASN at low levels, it is upregulated in cancers of the breast, prostate, and liver, among

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service