Skip to Content
Merck
All Photos(2)

Documents

SAB4300319

Sigma-Aldrich

Anti-MYC (Ab-358) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-bHLHe39 antibody produced in rabbit, Anti-c-Myc antibody produced in rabbit, Anti-v-myc myelocytomatosis viral oncogene homolog (avian) antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~60 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(R-R-T-H-N)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

General description

MYC (MYC proto-oncogene) is a transcriptional factor and oncoprotein. It is located on chromosome 8q24.

Immunogen

Peptide sequence around aa. 356-360 (R-R-T-H-N), according to the protein MYC.

Application

Anti-MYC (Ab-358) has been used in western blot analysis.

Biochem/physiol Actions

MYC (MYC proto-oncogene) may be essential for cancer initiation, promotion and therapy resistance. Overexpression of MYC in DLBCL (diffuse large B-cell lymphoma) results in poor outcome and invasive treatment when medicated with rituximab plus cyclophosphamide, doxorubicin, vincristine and prednisone (R-CHOP).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Myc proto-oncogene encodes nuclear DNA-binding phosphoproteins that are involved in the regulation of gene expression and DNA replication during cell growth and differentiation. Myc encodes a protein of 65 kDa which is expressed in almost all normal and transformed cells. The expression correlates with the proliferation state of the cells. Transcription is repressed in quiescent or terminally differentiated cells. Expression of Myc is generally induced after mitogenic stimulation of cells or serum induction. Myc therefore is an important positive regulator of cell growth and proliferation. Myc has been demonstrated also to be a potent inducer of apoptosis when expressed in the absence of serum or growth factors. Apoptosis may serve also as a protective mechanism to prevent tumorigenicity elicited by deregulated Myc expression. Sequences of the Myc oncogene have been highly conserved throughout evolution, from Drosophila to vertebrates.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

8q24 prostate, breast, and colon cancer risk loci show tissue-specific long-range interaction with MYC
Ahmadiyeh N, et al.
Proceedings of the National Academy of Sciences of the USA (2010)
Immortalized prairie vole-derived fibroblasts (VMF-K4DTs) can be transformed into pluripotent stem cells and provide a useful tool with which to determine optimal reprogramming conditions
Katayama M, et al,
Journal of Reproduction and Development (2017)
Voltage-dependent anion channel 1 interacts with ribonucleoprotein complexes to enhance infectious bursal disease virus polymerase activity
Han C, et al.
Journal of Virology (2017)
Clinicopathological significance of c-MYC in esophageal squamous cell carcinoma
Lian Y, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine (2017)
MYC Protein-positive Diffuse Large B-Cell Lymphoma Features an Activated B-Cell Receptor Signal Pathway
Wang WG, et al.
American Journal of Surgical Pathology (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service