Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300253

Sigma-Aldrich

Anti-phospho-SMAD3 (pSer425) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp586N0721 antibody produced in rabbit, Anti-DKFZp686J10186 antibody produced in rabbit, Anti-HSPC193 antibody produced in rabbit, Anti-HsT17436 antibody produced in rabbit, Anti-SMAD family member 3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~52 kDa

species reactivity

mouse, human, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100

isotype

IgG

immunogen sequence

(C-S-S-V-SP)

NCBI accession no.

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer425)

Gene Information

human ... SMAD3(4088)

General description

The SMAD3 (mothers against decapentaplegic homolog 3) gene is mapped to human chromosome 15q22.33. Smad3 protein identifies tandem repeats of the palindromic sequence GTCTAGAC, which is part of TGF-β (Transforming growth factor β) signaling promoter region.

Immunogen

Peptide sequence around phosphorylation site of serine 425 (C-S-S-V-S(p)), according to the protein SMAD3.

Application

Anti-phospho-SMAD3 (pSer425) antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Smad3 (mothers against decapentaplegic homolog 3) is associated with TGF-β (transforming growth factor β) signaling pathway, a cellular process regulating the homeostasis, development, and repair of cartilage. It is known to induce extracellular matrix production. Smad proteins are responsible for the transduction of signals from cell surface to the nucleus. Phosphorylation of Smad3 controls tumor progression, inflammation, fibrosis, obesity and diabetes. Smad3 expression might be associated with the pathogenesis of osteoarthritis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Smad3 encoded by this gene belongs to the SMAD, a family of proteins similar to the gene products of the Drosophila gene 'mothers against decapentaplegic' (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein functions as a transcriptional modulator activated by transforming growth factor-beta and is thought to play a role in the regulation of carcinogenesis.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Signaling via Smad2 and Smad3 is dispensable for adult murine hematopoietic stem cell function in vivo
Billing M, et al.
Experimental Hematology, 55, 34-44 (2017)
Tianpeng Xu et al.
Cytotechnology, 71(1), 57-65 (2019-01-02)
Mesenchymal stem cells (MSCs) hold great potential to treat tissue damage based on their multipotent property, and are also considered as suitable cell resources to create tissue-engineered grafts for tendon repair. However, the clinical application of MSCs is still limited
Histone deacetylase 3 unconventional splicing mediates endothelial-to-mesenchymal transition through transforming growth factor ?2
Zeng L, et al.
The Journal of Biological Chemistry, 288(44), 31853-31866 (2013)
Down-regulation of microRNA-216b inhibits IL-1β-induced chondrocyte injury by up-regulation of Smad3
He J, et al.
Bioscience Reports, 37(2) (2017)
Siu Chiu Chan et al.
Journal of the American Society of Nephrology : JASN, 29(10), 2493-2509 (2018-08-12)
Mutation of HNF1B, the gene encoding transcription factor HNF-1β, is one cause of autosomal dominant tubulointerstitial kidney disease, a syndrome characterized by tubular cysts, renal fibrosis, and progressive decline in renal function. HNF-1β has also been implicated in epithelial-mesenchymal transition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service