SAB4200706
Anti-TM4SF4 antibody, Mouse monoclonal
clone TM4SF-11, hybridoma cell culture supernatant
Synonym(s):
Anti-ILTMP, Anti-il-TMP transmembrane 4 L six family member 4
About This Item
Recommended Products
antibody form
culture supernatant
Quality Level
antibody product type
primary antibodies
clone
TM4SF-11, monoclonal
form
buffered aqueous solution
mol wt
antigen ~27 kDa
species reactivity
rat, human, canine, monkey, mouse, bovine
technique(s)
immunoblotting: 1:4,000-1:8,000 using whole extract of HCT-116 cells
isotype
IgM
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... TM4SF4(7104)
mouse ... Tm4sf4(229302)
rat ... Tm4sf4(116467)
General description
Immunogen
Application
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service