Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200622

Sigma-Aldrich

Anti-DISC-1 (Internal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody

Synonym(s):

C1orf136, DISC1, Disrupted in schizophrenia 1, LOC100039084, LOC640053, RP23−368K3.1, RP4−730B13.1, SCZD9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
species reactivity:
human, rat
application:
IP
technique(s):
immunoblotting: 1-2 μg/mL using whole extracts of rat brain P2 fraction or HEK-293T cells over-expressing human DISC1
immunoprecipitation (IP): 2.5-5.0 μg using extracts of mouse brain.
citations:
2

antibody form

affinity isolated antibody

Quality Level

antibody product type

primary antibodies

form

buffered aqueous solution

species reactivity

human, rat

concentration

~1 mg/mL

technique(s)

immunoblotting: 1-2 μg/mL using whole extracts of rat brain P2 fraction or HEK-293T cells over-expressing human DISC1
immunoprecipitation (IP): 2.5-5.0 μg using extracts of mouse brain.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DISC1(27185)
rat ... Disc1(307940)

Immunogen

synthetic peptide corresponding to an internal sequence in rat DISC1

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service