Skip to Content
Merck
All Photos(2)

Documents

SAB4200224

Sigma-Aldrich

Anti-phospho-TDP-43 (pSer410) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-phospho-ALS10, Anti-phospho-TAR DNA binding protein 43, Anti-phospho-TARDBP, Anti-phospho-TARDP43

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~43 kDa

species reactivity

mouse, human

concentration

~1.0 mg/mL

technique(s)

western blot: 1-2 μg/mL using HepG2 and NIH3T3 cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer410)

Gene Information

human ... TARDBP(23435)
mouse ... Tardbp(230908)

General description

TDP-43 (TAR DNA binding protein) belongs to the family of heterogeneous nuclear ribonucleoproteins (hnRNPs), that binds single stranded RNA. TDP-43 has been implicated in the transcription regulation of HIV. TDP-43 is the major ubiquinated component of cytoplasmic inclusions in frontotemporal lobe degeneration subtype FTLD-U and amyotrophic lateral sclerosis (ALS). TDP-43 is predominantly localized to the nucleus.

Immunogen

synthetic peptide containing phosphorylated Ser410 of human TDP-43, conjugated to KLH. The corresponding sequence is identical in mouse TDP-43.

Application

Anti-phospho-TDP-43 (pSer410) antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Pathological TDP-43 forms abnormal inclusions in neuronal perikarya and neurites. Several pathogenic TDP-43 mutations have been identified in familial amyotrophic lateral sclerosis (ALS).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margaret E Flanagan et al.
Journal of Alzheimer's disease : JAD, 66(4), 1549-1558 (2018-11-20)
Transactive response binding protein-43 (TDP-43) cytoplasmic neuronal and glial aggregates (pathologic TDP-43) have been described in multiple brain diseases. We describe the associations between neuropathologically confirmed TDP-43 and cognition in two population-based cohorts: the Nun Study (NS) and the Honolulu-Asia
Xiaoling Zhang et al.
Neuroscience bulletin, 35(2), 183-192 (2018-11-02)
Primary age-related tauopathy (PART) is characterized by tau neurofibrillary tangles (NFTs) in the absence of amyloid plaque pathology. In the present study, we analyzed the distribution patterns of phosphorylated 43-kDa TAR DNA-binding protein (pTDP-43) in the brains of patients with
Abnormal phosphorylation of Ser409/410 of TDP-43 in FTLD-U and ALS
Inukai Y, et al.
Febs Letters, 582(19), 2899-2904 (2008)
Peng Yin et al.
Acta neuropathologica, 137(6), 919-937 (2019-02-28)
The cytoplasmic accumulation of the nuclear TAR DNA-binding protein 43 (TDP-43) is a pathologic hallmark in amyotrophic lateral sclerosis, frontotemporal lobar degeneration, and other neurological disorders. However, most transgenic TDP-43 rodent models show predominant nuclear distribution of TDP-43 in the
Manuela Neumann et al.
Science (New York, N.Y.), 314(5796), 130-133 (2006-10-07)
Ubiquitin-positive, tau- and alpha-synuclein-negative inclusions are hallmarks of frontotemporal lobar degeneration with ubiquitin-positive inclusions and amyotrophic lateral sclerosis. Although the identity of the ubiquitinated protein specific to either disorder was unknown, we showed that TDP-43 is the major disease protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service