Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500598

Sigma-Aldrich

Anti-ZIP8 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-BIGM103, Anti-PP3105, Anti-SLC39A8, Anti-Slc39A8, Anti-Solute carrier family 39 member A8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC39A8(64116)

General description

Solute carrier family 39 member 8 (SLC39A8), also known as zinc- and iron-related protein 8 (ZIP8), is a plasma membrane transporter protein. It is part of the solute carrier gene superfamily. The gene encoding SLC39A8 is localized on human chromosome 4q24.

Immunogen

ZIP8 antibody was raised against a 18 amino acid peptide near the carboxy terminus of human ZIP8.

Application

Anti-ZIP8 antibody produced in rabbit has been used in immunocytochemistry.

Biochem/physiol Actions

Solute carrier family 39 member 8 (SLC39A8) co-transports divalent metal cations like cadmium, zinc, manganese and iron. It has a crucial role in the cellular uptake of cadmium. The protein is a target of nuclear-factor κB (NF-κB) and has a role in pro-inflammatory responses.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500598.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cellular localization and developmental
changes of Zip8, Zip14 and transferrin
receptor 1 in the inner ear of rats
Dalian Ding
Biometals: An International Journal on the Role of Metal Ions in Biology, Biochemistry, And Medicine (2014)
ZIP8 regulates host defense through zinc-mediated inhibition of NF-?B.
Liu MJ
Cell Reports (2013)
A blood pressure-associated variant of the SLC39A8 gene influences cellular cadmium accumulation and toxicity.
Zhang R
Human Molecular Genetics (2016)
Advances in the genetics of schizophrenia: toward a network and pathway view for drug discovery: Advances in the genetics of schizophrenia
David A
The New York State Dental Journal (2016)
Autosomal-Recessive Intellectual Disability with Cerebellar Atrophy Syndrome Caused by Mutation of the Manganese and Zinc Transporter Gene SLC39A8.
Boycott KM
American Journal of Human Genetics (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service