Skip to Content
Merck
All Photos(10)

Key Documents

SAB2701959

Sigma-Aldrich

Anti-HMGA2 antibody produced in rabbit

Synonym(s):

HMGI-C, HMGIC, LIPO, STQTL9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

12 kDa

species reactivity

mouse, human, rat

concentration

0.7 mg/mL

technique(s)

ChIP: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: 5000-50000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HMGA2(8091)

General description

High mobility group A2 (HMGA2) protein is a non-histone architectural transcription factor. This protein belongs to the high-mobility group adenine/thymine (AT)-hook (HMGA) family. HMGA2 is expressed in the mesenchyme prior differentiation and tumors of epithelial origin. The HMGA2 gene is located on the human chromosome at 12q14.3.

Immunogen

Recombinant fragment corresponding to a region within amino acids 1 and 109 of HMGA2 (Uniprot ID#P52926)

Application

Anti-HMGA2 antibody produced in rabbit has been used in immunohistochemistry (1:250), (1:100), (1:4000).

Biochem/physiol Actions

High mobility group A2 (HMGA2) protein is involved in regulating the activity of various genes by binding to the adenine/thymine (AT)-rich sequences present in the minor groove of B‑form DNA. This protein plays a role in many biological processes such as the DNA damage repair process, senescence, apoptosis, and cell cycle process. HMGA2 protein also plays a role in epithelial-mesenchymal transition and telomere restoration. Overexpression of HMG2A gene is observed in metastatic lung adenocarcinoma.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madhu S Kumar et al.
Nature, 505(7482), 212-217 (2013-12-07)
Non-small-cell lung cancer (NSCLC) is the most prevalent histological cancer subtype worldwide. As the majority of patients present with invasive, metastatic disease, it is vital to understand the basis for lung cancer progression. Hmga2 is highly expressed in metastatic lung
Yong Sun Lee et al.
Genes & development, 21(9), 1025-1030 (2007-04-18)
HMGA2, a high-mobility group protein, is oncogenic in a variety of tumors, including benign mesenchymal tumors and lung cancers. Knockdown of Dicer in HeLa cells revealed that the HMGA2 gene is transcriptionally active, but its mRNA is destabilized in the
Shizhen Zhang et al.
International journal of oncology, 55(4), 775-788 (2019-08-23)
The high mobility group A2 (HMGA2) protein is a non‑histone architectural transcription factor that modulates the transcription of several genes by binding to AT‑rich sequences in the minor groove of B‑form DNA and alters the chromatin structure. As a result, HMGA2
Zheng Hu et al.
Nature genetics, 47(2), 158-163 (2015-01-13)
Human papillomavirus (HPV) integration is a key genetic event in cervical carcinogenesis. By conducting whole-genome sequencing and high-throughput viral integration detection, we identified 3,667 HPV integration breakpoints in 26 cervical intraepithelial neoplasias, 104 cervical carcinomas and five cell lines. Beyond
Asahiro Morishita et al.
Cancer research, 73(14), 4289-4299 (2013-06-01)
The non-histone chromatin-binding protein HMGA2 is expressed predominantly in the mesenchyme before its differentiation, but it is also expressed in tumors of epithelial origin. Ectopic expression of HMGA2 in epithelial cells induces epithelial-mesenchymal transition (EMT), which has been implicated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service